Skip to Content
Merck
All Photos(1)

Key Documents

D2205000

Dimetindene maleate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N,N-Dimethyl-2-{3-[(RS)-1-(pyridine-2-yl)ethyl]-1H-inden-2-yl}ethanamin-(Z)-butendioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24N2 · C4H4O4
CAS Number:
Molecular Weight:
408.49
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

dimetindene

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

N(CCC1=C(c3c(cccc3)C1)C(C)c2ncccc2)(C)C.OC(=O)\C=C/C(=O)O

InChI

1S/C20H24N2.C4H4O4/c1-15(19-10-6-7-12-21-19)20-17(11-13-22(2)3)14-16-8-4-5-9-18(16)20;5-3(6)1-2-4(7)8/h4-10,12,15H,11,13-14H2,1-3H3;1-2H,(H,5,6)(H,7,8)/b;2-1-

InChI key

SWECWXGUJQLXJF-BTJKTKAUSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dimetindene maleate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Immunoglobulin E-mediated anaphylaxis to rabeprazole.
V Vovolis et al.
Journal of investigational allergology & clinical immunology, 20(4), 360-361 (2010-09-08)
Urticaria, angioedema and dyspnoea in adjuvant therapy of melanoma with interferon alpha-2b.
Doris Helbig et al.
Acta dermato-venereologica, 90(3), 308-309 (2010-06-09)
Christos Kosmas et al.
Medical science monitor : international medical journal of experimental and clinical research, 12(11), CR462-CR466 (2006-10-31)
Many investigations have focused on optimal doses/schedules since regulatory agency approval of paclitaxel (Taxol). Paclitaxel is generally administered at doses of 175-225 mg/m2 over 3 hours or 135-175 mg/m2 over 24 hours, every 3 weeks. The purpose of this study
C Dopfer et al.
Klinische Padiatrie, 222(7), 460-461 (2010-09-11)
We report a case of acute hemorrhagic edema in a 10-year-old girl caused by adder bite. Therapy consisted of analgesics, antihistamines, corticosteroids. Antivenum serum was applied in a single dose due to severe spreading of the edema. The patient recovered
[Aso an inspection of the family dog is warrented here].
M F Nentwich et al.
MMW Fortschritte der Medizin, 148(42), 5-5 (2007-07-12)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service