Skip to Content
Merck
All Photos(1)

Key Documents

D0730000

Dexpanthenol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

D-Panthenol, (R)-(+)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutyramide, (R)-2,4-Dihydroxy-3,3-dimethylbutyric 3-hydroxypropylamide, D-Pantothenyl alcohol, Dexpanthenol, Provitamin B

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H19NO4
CAS Number:
Molecular Weight:
205.25
Beilstein:
1724947
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

biological source

synthetic

grade

pharmaceutical primary standard

Agency

EP Reference Standard

API family

dexpanthenol

form

solid

manufacturer/tradename

EDQM

technique(s)

gas chromatography (GC): suitable
liquid chromatography (LC): suitable

density

1.20 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC(C)(CO)[C@@H](O)C(=O)NCCCO

InChI

1S/C9H19NO4/c1-9(2,6-12)7(13)8(14)10-4-3-5-11/h7,11-13H,3-6H2,1-2H3,(H,10,14)/t7-/m0/s1

InChI key

SNPLKNRPJHDVJA-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.
Dexpanthenol, an alcoholic analog of pantothenic acid, can be potentially applied for skin wound healing application. It is utilized in multi-vitamin formulations, as a parenteral drug and certain cosmetic preparations. When absorbed by the skin, it is transformed into pantothenic acid (vitamin B5), which has implications in the biosynthesis of coenzyme A.

Application

This European Pharmacopoeia reference standard is intended for use only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Simultaneous quantification of dexpanthenol and resorcinol from hair care formulation using liquid chromatography: method development and validation
De, Amit Kumar and Chowdhury, Partha Pratim and Chattapadhyay, Shyamaprasad
Scientifica, 2016(3), 983-988 (2016)
Simultaneous determination of mepyramine maleate, lidocaine hydrochloride, and dexpanthenol in pharmaceutical preparations by partial least-squares multivariate calibration
Aksu O, et al.
Analytical Letters, 39(4), 751-761 (2006)
Anti-inflammatory and anti-oxidative effects of dexpanthenol on lipopolysaccharide induced acute lung injury in mice
Li-Mei W, et al.
Inflammation, 39(5), 1757-1763 (2016)
Dexpanthenol
European Pharmacopoeia Commission and European Directorate for the Quality of Medicines & Healthcare
European pharmacopoeia, 2016(3), 2227-2228 (2017)
Development and validation of reversed phase high performance liquid chromatography method for determination of dexpanthenol in pharmaceutical formulations.
Kulikov U A and Zinchenko A A.
Journal of Pharmaceutical and Biomedical Analysis, 43(3), 983-988 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service