Skip to Content
Merck
All Photos(3)

Key Documents

C1000

Sigma-Aldrich

Calcium acetate hydrate

ReagentPlus®, ≥99% (titration), powder

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ca · xH2O
CAS Number:
Molecular Weight:
158.17 (anhydrous basis)
Beilstein:
3692527
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.31

Quality Level

product line

ReagentPlus®

Assay

≥99% (titration)

form

powder

color

white

solubility

water: 200 mg/mL, clear to slightly hazy, colorless to faintly yellow

SMILES string

O.CC(=O)O[Ca]OC(C)=O

InChI

1S/2C2H4O2.Ca.H2O/c2*1-2(3)4;;/h2*1H3,(H,3,4);;1H2/q;;+2;/p-2

InChI key

XQKKWWCELHKGKB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application


  • Combined experimental and theoretical studies on adsorption mechanisms of gaseous mercury(II) by calcium-based sorbents: The effect of unsaturated oxygen sites.: This research investigates the adsorption mechanisms of mercury(II) using calcium-based sorbents, emphasizing the environmental applications of calcium acetate hydrate (Tang et al., 2019).

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Frick et al.
Molecular cancer research : MCR, 16(4), 634-642 (2018-01-31)
Patients with inflammatory bowel disease (IBD) have a higher risk of developing colitis-associated-cancer (CAC); however, the underlying processes of disease progression are not completely understood. Here, the molecular processes of inflammation-driven colon carcinogenesis were investigated using IL10-deficient mice (IL10 KO).
Eun A Ko et al.
American journal of physiology. Cell physiology, 304(11), C1042-C1052 (2013-02-22)
Electromechanical coupling via membrane depolarization-mediated activation of voltage-dependent Ca(2+) channels (VDCC) is an important mechanism in regulating pulmonary vascular tone, while mouse is an animal model often used to study pathogenic mechanisms of pulmonary vascular disease. The function of VDCC
Anamarija Markota et al.
PloS one, 14(7), e0218332-e0218332 (2019-07-06)
Clinical observations in inflammatory bowel disease patients and experimental studies in rodents suggest that iron in the intestinal lumen derived from iron-rich food or oral iron supplementation could exacerbate inflammation and that iron depletion from the diet could be protective.
Un Seng Chio et al.
Cell reports, 26(1), 37-44 (2019-01-04)
Molecular chaperones play key roles in maintaining cellular proteostasis. In addition to preventing client aggregation, chaperones often relay substrates within a network while preventing off-pathway chaperones from accessing the substrate. Here we show that a conserved lid motif lining the
Dilek Teker et al.
Materials science & engineering. C, Materials for biological applications, 48, 579-585 (2015-01-13)
An innovative multi-layer coating comprising a bioactive compound layer (consisting of hydroxyapatite and calcium titanate) with an underlying titanium oxide layer (in the form of anatase and rutile) has been developed on Grade 4 quality commercially pure titanium via a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service