Skip to Content
Merck
All Photos(1)

Key Documents

B1390

Supelco

Bisacodyl

analytical standard, for drug analysis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H19NO4
CAS Number:
Molecular Weight:
361.39
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard, for drug analysis

Quality Level

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)
veterinary

format

neat

storage temp.

2-8°C

SMILES string

CC(=O)Oc1ccc(cc1)C(c2ccc(OC(C)=O)cc2)c3ccccn3

InChI

1S/C22H19NO4/c1-15(24)26-19-10-6-17(7-11-19)22(21-5-3-4-14-23-21)18-8-12-20(13-9-18)27-16(2)25/h3-14,22H,1-2H3

InChI key

KHOITXIGCFIULA-UHFFFAOYSA-N

Gene Information

human ... CHRM4(1132)

Looking for similar products? Visit Product Comparison Guide

General description

Bisacodyl is a diphenylmethane derivative, mainly used as a stimulant laxative for treating constipation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael A Kamm et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 9(7), 577-583 (2011-03-29)
Although stimulant laxatives have been used for many years to treat patients with constipation, their clinical value has been questioned, and there have been few high-quality trials to assess their efficacy. We tested the efficacy and safety of 4 weeks
V Giorgio et al.
Neurogastroenterology and motility : the official journal of the European Gastrointestinal Motility Society, 25(1), 70-78 (2012-10-04)
Severe pediatric slow transit constipation (STC) is commonly due to intrinsic colonic neuromuscular disease. We sought to correlate neuromuscular histological phenotypes in pediatric STC with colonic manometric phenotypes using high-resolution manometry (HRM). We tested the hypothesis that failure of motor
Drugs in Pregnancy and Lactation: A Reference Guide to Fetal and Neonatal Risk (2011)
Mercedes Ibáñez et al.
Diseases of the colon and rectum, 54(12), 1578-1584 (2011-11-10)
No consensus exists regarding the optimal bowel preparation regimen for patients with poor bowel cleansing at a previous colonoscopy. We investigated the usefulness of an intensive cleansing regimen for repeat colonoscopy after previous failure of bowel preparation. A prospective observational
Christian Friedrich et al.
Drug metabolism and pharmacokinetics, 26(5), 458-464 (2011-06-24)
The aim of this study was to determine whether administration of the prodrugs bisacodyl (Bisa) and sodium picosulfate (SPS) leads to excretion of their common active metabolite, bis-(p-hydroxyphenyl)-pyridyl-2-methane (BHPM), in breast milk. Two groups of 8 healthy lactating women who

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service