Skip to Content
Merck
All Photos(1)

Key Documents

B1152050

Bromperidol decanoate

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Decanoic acid 4-(4-bromophenyl)-1-[4-(4-fluorophenyl)-4-oxobutyl]-4-piperidinyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H41BrFNO3
CAS Number:
Molecular Weight:
574.56
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

bromperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC(CC2)(OC(=O)CCCCCCCCC)c3ccc(cc3)Br

InChI

1S/C31H41BrFNO3/c1-2-3-4-5-6-7-8-11-30(36)37-31(26-14-16-27(32)17-15-26)20-23-34(24-21-31)22-9-10-29(35)25-12-18-28(33)19-13-25/h12-19H,2-11,20-24H2,1H3

InChI key

ZINCPWWBSRSXBH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Bromperidol decanoate EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Smeraldi et al.
Minerva psichiatrica, 37(1), 39-44 (1996-03-01)
We assessed the safety and therapeutic efficacy of bromperidol decanoate in a six-month open trial on 21 patients (13 males and 8 females), mean age 41.3 years, who were no longer in the acute phase and were being treated as
S Quraishi et al.
The Cochrane database of systematic reviews, (2)(2), CD001719-CD001719 (2000-05-05)
Anti-psychotic drugs are the mainstay treatment for schizophrenia. Long-acting depot injections of drugs such as bromperidol decanoate are extensively used as a means of long-term maintenance treatment. To assess the effects of depot bromperidol versus placebo, oral anti-psychotics and other
Marianna Purgato et al.
The Cochrane database of systematic reviews, (9)(9), CD001719-CD001719 (2011-09-09)
Antipsychotic drugs are the mainstay treatment for schizophrenia. Long-acting depot injections of drugs such as bromperidol decanoate are extensively used as a means of long-term maintenance treatment. To assess the effects of depot bromperidol versus placebo, oral antipsychotics and other
Marianna Purgato et al.
The Cochrane database of systematic reviews, 11, CD001719-CD001719 (2012-11-16)
Antipsychotic drugs are the mainstay treatment for schizophrenia. Long-acting depot injections of drugs such as bromperidol decanoate are extensively used as a means of long-term maintenance treatment. To assess the effects of depot bromperidol versus placebo, oral antipsychotics and other
A Levi Minzi et al.
Minerva psichiatrica, 33(1), 51-55 (1992-01-01)
An open study in 20 patients (7 M, 13 F, mean age 40 years) has been performed in order to evaluate the effectiveness and the safety of bromperidol decanoate in the long-term treatment of psychotic disorders. Patients were selected according

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service