Skip to Content
Merck
All Photos(1)

Key Documents

B0470000

Benperidol

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

1-{1-[4-(4-Fluorophenyl)-4-oxobutyl]-piperidin-4-yl}-1,3-dihydro-benzimidazol-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H24FN3O2
CAS Number:
Molecular Weight:
381.44
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

benperidol

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)C(=O)CCCN2CCC(CC2)[n]3c4c([nH][c]3=O)cccc4

InChI

1S/C22H24FN3O2/c23-17-9-7-16(8-10-17)21(27)6-3-13-25-14-11-18(12-15-25)26-20-5-2-1-4-19(20)24-22(26)28/h1-2,4-5,7-10,18H,3,6,11-15H2,(H,24,28)

InChI key

FEBOTPHFXYHVPL-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Benperidol EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jo Ann V Antenor-Dorsey et al.
European journal of nuclear medicine and molecular imaging, 35(4), 771-778 (2007-12-12)
N-([(11)C]methyl)benperidol ([(11)C]NMB) can be used for positron emission tomography (PET) measurements of D(2)-like dopamine receptor binding in vivo. We report the absorbed radiation dosimetry of i.v.-administered (11)C-NMB, a critical step before applying this radioligand to imaging studies in humans. Whole-body
S Leucht et al.
The Cochrane database of systematic reviews, (1)(1), CD003083-CD003083 (2002-03-01)
Benperidol is a relatively old antipsychotic drug, marketed since 1966, used in Germany for 30 years, but also available in Belgium, Greece, Italy, the Netherlands and the UK. Benperidol is a butyrophenone antipsychotic, with the highest neuroleptic potency in terms
Complete dopamine D2 receptor occupancy without extrapyramidal side effects under benperidol.
Carlos Schönfeldt-Lecuona et al.
Journal of clinical psychopharmacology, 24(1), 97-98 (2004-01-08)
J Schröder et al.
Psychiatry research, 75(2), 115-123 (1997-11-14)
Recent studies have described a left lateralized striatal asymmetry of D2 dopamine receptors in male patients with schizophrenia. To replicate this finding and to explore its potential functional consequences, we investigated the D2 dopamine receptor system in 23 drug-naive patients
J Schröder et al.
Fortschritte der Neurologie-Psychiatrie, 68 Suppl 1, S42-S45 (2000-07-25)
Animal and post mortem studies indicate that neuroleptic therapy may induce D2-dopamine receptor upregulation in the basal ganglia. To address this phenomenon in a clinical study, we investigated the D2-dopamine receptor binding in 15 DSM-III-R schizophrenics in the drug-naive state

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service