Skip to Content
Merck
All Photos(5)

Key Documents

B0149

Sigma-Aldrich

Brilliant Blue R

Dye content ~50 %, Powder

Synonym(s):

Acid Blue 83, Brilliant indocyanin 6B, Coomassie Brilliant Blue R

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C45H44N3NaO7S2
CAS Number:
Molecular Weight:
825.97
Colour Index Number:
42660
Beilstein:
5718025
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

product name

Brilliant Blue R, Dye content ~50 %, Technical grade

Quality Level

form

powder

composition

Dye content, ~50%

solubility

H2O: 10 mg/mL

suitability

suitable for detection of protein bands following electrophoresis.

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Na+].CCOc1ccc(Nc2ccc(cc2)C(\c3ccc(cc3)N(CC)Cc4cccc(c4)S([O-])(=O)=O)=C5\C=C/C(C=C5)=[N+](\CC)Cc6cccc(c6)S([O-])(=O)=O)cc1

InChI

1S/C45H45N3O7S2.Na/c1-4-47(31-33-9-7-11-43(29-33)56(49,50)51)40-23-15-36(16-24-40)45(35-13-19-38(20-14-35)46-39-21-27-42(28-22-39)55-6-3)37-17-25-41(26-18-37)48(5-2)32-34-10-8-12-44(30-34)57(52,53)54;/h7-30H,4-6,31-32H2,1-3H3,(H2,49,50,51,52,53,54);/q;+1/p-1

InChI key

NKLPQNGYXWVELD-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Brilliant Blue R (C.l. 42660) is a triaminotriarylmethane dye with two amino groups being benzylated and one phenylated. It is also known as brilliant indocyanine 6B and coomassie blue R250.

Application

Brilliant Blue R has been widely used for protein band detection after SDS-PAGE (sodium dodecyl sulfate-polyacrylamide gel electrophoresis).Its other applications include:
  • as a protein stain on epoxy-embedded tissue sections
  • for quantitative micro densitometry of protein in sections
  • for staining chromatids, actinin microfilaments, developing neurites, and sperm acrosomes.
For detection of protein bands following electrophoresis.

Legal Information

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Role of matrix metalloproteinase-2 in the cardioprotective effect of ischaemic postconditioning.
Donato M, et al.
Experimental Physiology, 95, 274-281 (2010)
Effect of Flammulina velutipes on spent-hen breast meat tenderization.
Kang GH, et al.
Poultry Science, 91, 232-236 (2012)
Rozanne Arulanandam et al.
Communications biology, 3(1), 254-254 (2020-05-24)
We have demonstrated that microtubule destabilizing agents (MDAs) can sensitize tumors to oncolytic vesicular stomatitis virus (VSVΔ51) in various preclinical models of cancer. The clinically approved T-DM1 (Kadcyla®) is an antibody-drug conjugate consisting of HER2-targeting trastuzumab linked to the potent
Marina Piscopo et al.
Hearing research, 178(1-2), 89-94 (2003-04-10)
The factors controlling otoconia growth are not well known but it seems that the type of proteins contained in the otoconia regulates the initiation and/or the subsequent rates of crystal growth determining the morphology and the size of the final
Grit Nebrich et al.
Electrophoresis, 28(10), 1607-1614 (2007-04-21)
The availability of easy-to-handle, sensitive, and cost-effective protein staining protocols for 2-DE, in conjunction with a high compatibility for subsequent MS analysis, is still a prerequisite for successful proteome research. In this article we describe a quick and easy-to-use methodological

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service