Skip to Content
Merck
All Photos(1)

Key Documents

99311

Supelco

Potassium ionophore I - cocktail A

Selectophore

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NB.61

product line

Selectophore

Quality Level

form

liquid

composition

Dibutyl sebacate, 68.0 wt. % (84838)
1,2-Dimethyl-3-nitrobenzene, 25.0 wt. % (40870)
Potassium ionophore I, 5.0 wt. % (60403)
Potassium tetrakis(4-chlorophenyl)borate, 2.0 wt. % (60591)

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Clear borosilicate glass, high hydrolytic resistance (Type I)

Other Notes

Ready-to-use ion selective liquid for the preparation of microelectrodes in micropipettes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D. Ammann et al.
Ion-Selective Microelectrodes and their Use in Excitable Tissues, 13-13 (1981)
E.Sykova et al.,eds.
Ion-Selective Microelectrodes and their Use in Excitable Tissues, Plenum Press null
Mengliang Niu et al.
Journal of experimental botany, 69(20), 4945-4960 (2018-07-12)
Tissue tolerance to salinity stress is a complex physiological trait composed of multiple 'sub-traits' such as Na+ compartmentalization, K+ retention, and osmotic tolerance. Previous studies have shown that some Cucurbita species employ tissue tolerance to combat salinity and we aimed
D. Ammann et al.
Ion-Selective Microelectrodes and their Use in Excitable Tissues, 13 -13 (1981)
Giselle Cheung et al.
The Journal of physiology, 587(Pt 4), 753-768 (2008-12-03)
Microglial cells (brain macrophages) invade the brain during embryonic and early postnatal development, migrate preferentially along fibre tracts to their final position and transform from an amoeboid to a ramified morphology. Signals by which the invading microglia communicate with other

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service