Skip to Content
Merck
All Photos(1)

Key Documents

96351

Supelco

Pullulan Standard Set Mp ~350 - 700′000

set of analytical standards, for GPC

Synonym(s):

Pullulan Standard Set

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41116107
E Number:
E1204
NACRES:
NA.24

grade

analytical standard
for GPC

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

oligosaccharides

technique(s)

gel permeation chromatography (GPC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Pullulan is a linear glucan polysaccharide made up of α-1,4-maltotriose repeating units. It is produced by yeast-like fungus Aureobasidium pullulans from starch and has α-1,6 linkages between two maltotriose units.

Application

The set of analytical standards can also be used as follows:
  • Calibration of the polymer-based column in the estimation of molecular weight of the polysaccharide fractions obtained from seaweeds— S. latissima and M. stellatus, by a high-performance size exclusion chromatography (HPSEC) method
  • Measurement of molecular masses of oligomeric cellulose by size exclusion chromatography in combination with multi-angle laser light scattering (MALLS) detection
  • To calibrate the SEC column in the determination of molecular size distribution of microalgal native glycogen
  • Quantitative analysis of membrane formation process of alginate–chitosan microcapsules using gel permeation chromatography(GPC).

Components

Kit with carefully selected reference materials to calibrate GPC/SEC/GFC systems. The kit contains standards of the same type, but with different molar masses, a certificate of analysis (CoA) for each standard and an example calibration curve with analytical conditions.
The standards need to be weighed before use. Suitable for calibrations in aqueous solutions and DMSO.
Kit contains 10 different pullulan standards, each 100 mg, with Mp ranging from ~350 to ~700,000.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Quantitative characterization of membrane formation process of alginate?chitosan microcapsules by GPC
Yu W, et al.
Journal of Membrane Science, 346, 296-301 (2010)
Pei-Hong Miao et al.
Die Pharmazie, 67(3), 229-232 (2012-04-26)
Current efforts had been made to undertake a three-dimensional (3-D) reverse transfection of bone marrow-derived mesenchymal stem cells (BM-MSCs) in PLGA scaffolds. As a kind of multipotent stem cells, BM-MSCs show great potential and tremendous capacity in the gene transfection
Mélanie Lavergne et al.
Macromolecular bioscience, 12(7), 901-910 (2012-06-15)
Human ECFCs contribute to vascular repair. For this reason, they are considered as valuable cell therapy products in ischemic diseases. Porous scaffolds are prepared that are composed of natural polysaccharides, pullulan and dextran, by chemical crosslinking without use of organic
Rahul Garhwal et al.
Investigative ophthalmology & visual science, 53(3), 1341-1352 (2012-01-24)
To formulate conventional contact lenses that incorporate nanosphere-encapsulated antibiotic and demonstrate that the lenses provide for sustained antibacterial activity. A copolymer composed of pullulan and polycaprolactone (PCL) was used to synthesize core-shell nanospheres that encapsulated ciprofloxacin. Bactericidal activity of the
Marieta Constantin et al.
Carbohydrate polymers, 91(1), 74-84 (2012-10-10)
Pullulan-graft-poly(3-acrylamidopropyl trimethylammonium chloride) (P-g-pAPTAC) microspheres were prepared by suspension cross-linking of the pullulan previously grafted with cationic moieties. Adsorption of Azocarmine B by the P-g-pAPTAC microspheres was used as a model to demonstrate the removal of anionic dyes from aqueous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service