Skip to Content
Merck
All Photos(1)

Key Documents

95381

Sigma-Aldrich

Bismuth(III) oxide

purum, ≥98.0% (KT)

Synonym(s):

Dibismuth trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Bi2O3
CAS Number:
Molecular Weight:
465.96
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

grade

purum

Quality Level

Assay

≥98.0% (KT)

form

crystals

reaction suitability

reagent type: catalyst
core: bismuth

loss

≤0.2% loss on ignition

SMILES string

O=[Bi]O[Bi]=O

InChI

1S/2Bi.3O

InChI key

WMWLMWRWZQELOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

Preparation of new stabilized, oxide ion-conducting, bismuth vanadate phases by a microwave assisted method, from V2O5, Bi2O3 and other solid oxides, was reported. These ceramics show promise in solid oxide fuel cells, water-vapor electrolyzers and oxygen sensors.

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lu You et al.
ACS nano, 6(6), 5388-5394 (2012-05-18)
The novel strain-driven morphotropic phase boundary (MPB) in highly strained BiFeO(3) thin films is characterized by well-ordered mixed phase nanodomains (MPNs). Through scanning probe microscopy and synchrotron X-ray diffraction, eight structural variants of the MPNs are identified. Detailed polarization configurations
Rifath Sharmin et al.
Journal of the Air & Waste Management Association (1995), 62(9), 1032-1039 (2012-10-02)
Photocatalytic oxidation (PCO) is a promising technology for indoor air purification due to low operating cost, potentially long service life, and low maintenance. Ultraviolet light-emitting diode (UVLED) is a new concept in the field of PCO, which has several advantages
L M Formosa et al.
International endodontic journal, 45(4), 326-336 (2011-11-03)
To investigate the physical properties of tricalcium silicate (TCS) with and without the addition of a radiopacifier and compare them with that of Portland cement (PC) and radiopaque PC in an mineral trioxide aggregate-like system. Tricalcium silicate, PC and radiopacified
V S Kopp et al.
Acta crystallographica. Section A, Foundations of crystallography, 68(Pt 1), 148-155 (2011-12-22)
X-ray diffraction from films consisting of layers with different thicknesses, structures and chemical contents is analysed. The disorder is described by probabilities for different sequences of layers. Closed analytical expressions for the diffracted X-ray intensity are obtained when the layers
Luonothar Antunes Schmitt Dreger et al.
Journal of endodontics, 38(3), 324-329 (2012-02-22)
Mineral trioxide aggregate (MTA) and Portland cement have been shown to be bioactive because of their ability to produce biologically compatible carbonated apatite. This study analyzed the interaction of MTA and white Portland cement with dentin in vivo. Seventy-two human dentin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service