Skip to Content
Merck
All Photos(1)

Documents

92136

Supelco

Genistein

analytical standard

Synonym(s):

4′,5,7-Trihydroxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-4H-1-benzopyran-4-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10O5
CAS Number:
Molecular Weight:
270.24
Beilstein:
263823
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

Oc1ccc(cc1)C2=COc3cc(O)cc(O)c3C2=O

InChI

1S/C15H10O5/c16-9-3-1-8(2-4-9)11-7-20-13-6-10(17)5-12(18)14(13)15(11)19/h1-7,16-18H

InChI key

TZBJGXHYKVUXJN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Genistein, a naturally occurring soy isoflavone, is a dietary protein tyrosine kinase (PTK) inhibitor. It is found to possess anticarcinogenic properties.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Inhibitor of tyrosine protein kinase; competitive inhibitor of ATP in other protein kinase reactions. Antiangiogenic agent, down-regulates the transcription of genes involved in controlling angiogenesis.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: glycyrrhiza

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genistein inhibits both estrogen and growth factor-stimulated proliferation of human breast cancer cells
GP and SB
Cell Growth & Differentiation : the Molecular Biology Journal of the American Association For Cancer Research, 7, 1345-1351 (1996)
Alexandra Mamagkaki et al.
PloS one, 16(4), e0250599-e0250599 (2021-04-28)
The objective of this study is to improve and optimize the formulation of Genistein in capsules in order to result in a better pharmacokinetic profile comparing to existing commercial products. In order to do this, five different formulations of Genistein
Genistein inhibits renal cancer progression through long non-coding RNA HOTAIR suppression
Imai-Sumida M, et al.
Clinical Cancer Research, 3449-3449 (2017)
Sonia de Pascual-Teresa et al.
The Journal of nutritional biochemistry, 17(4), 257-264 (2005-08-20)
If soy isoflavones are to be effective in preventing or treating a range of diseases, they must be bioavailable, and thus understanding factors which may alter their bioavailability needs to be elucidated. However, to date there is little information on
M S Morton et al.
The Journal of endocrinology, 142(2), 251-259 (1994-08-01)
Plasma levels of the lignans enterodiol and enterolactone, and also the isoflavonic phyto-oestrogens daidzein, equol and genistein, are reported for postmenopausal Australian women consuming a traditional diet supplemented with linseed, soya flour or clover sprouts. Analysis was performed by gas

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service