Skip to Content
Merck
All Photos(1)

Documents

91988

Supelco

Tridecanoic acid

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)11CO2H
CAS Number:
Molecular Weight:
214.34
Beilstein:
508317
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

236 °C/100 mmHg (lit.)

mp

41-42 °C (lit.)
42-44 °C

format

neat

functional group

carboxylic acid

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCC(O)=O

InChI

1S/C13H26O2/c1-2-3-4-5-6-7-8-9-10-11-12-13(14)15/h2-12H2,1H3,(H,14,15)

InChI key

SZHOJFHSIKHZHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tridecanoic acid may be used as an internal standard for the :
  • analysis of pentaerythritol, phthalic acid and triglycerides in extracts from industrial alkyd resins using gas chromatography and high-performance liquid chromatography coupled to mass spectrometry
  • quantification of metabolites of arachidonic acid from cultures of endothelial cells using high performance liquid chromatography coupled to mass spectrometry

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Accelerated triacylglycerol production and altered fatty acid composition in oleaginous microalga Neochloris oleoabundans by overexpression of diacylglycerol acyltransferase 2
Klaitong P, et al.
Microbial cell factories, 16, 61-61 (2017)
Identification and quantification of metabolites of arachidonic acid from cultures of endothelial cells by HPLC-MS2
Zelinski.V and Borlak.J
Xenobiotica, 35(8), 811-823 (2015)
Industrial alkyd resins: characterization of pentaerythritol and phthalic acid esters using integrated mass spectrometry
Nasa LJ, et al.
Rapid Communications in Mass Spectrometry, 29(3), 1232-1241 (2015)
Hongchao Gou et al.
Frontiers in cellular and infection microbiology, 10, 599087-599087 (2021-02-16)
For viral replication to occur in host cells, low-molecular-weight metabolites are necessary for virion assembly. Recently, metabolomics has shown great promise in uncovering the highly complex mechanisms associated with virus-host interactions. In this study, the metabolic networks in PK-15 cells
Xiaohong Yang et al.
Marine drugs, 17(4) (2019-04-03)
Marine cyanobacteria represent a large untapped source of functional glycolipids enriched with polyunsaturated fatty acids (PUFAs) for human health. However, advanced methods for scalable isolation of diverse species containing high-purity PUFA-rich glycolipids will have to be developed and their possible

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service