Skip to Content
Merck
All Photos(1)

Documents

90669

Sigma-Aldrich

(±)-α-Tocopherol

tested according to Ph. Eur.

Synonym(s):

all-rac-α-Tocopherolum, DL-all-rac-α-Tocopherol, Vitamin E

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H50O2
CAS Number:
Molecular Weight:
430.71
Beilstein:
94012
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
34058016
PubChem Substance ID:
NACRES:
NA.26

biological source

synthetic

Quality Level

Agency

USP/NF
tested according to Ph. Eur.

Assay

96.0-102.0%

form

liquid

color

tan

mp

2.5-3.5 °C

density

0.950 g/mL at 20 °C (lit.)

application(s)

pharmaceutical (small molecule)

storage temp.

2-8°C

SMILES string

CC(C)CCCC(C)CCCC(C)CCCC1(C)CCc2c(C)c(O)c(C)c(C)c2O1

InChI

1S/C29H50O2/c1-20(2)12-9-13-21(3)14-10-15-22(4)16-11-18-29(8)19-17-26-25(7)27(30)23(5)24(6)28(26)31-29/h20-22,30H,9-19H2,1-8H3

InChI key

GVJHHUAWPYXKBD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kirsten G Coupland et al.
Movement disorders : official journal of the Movement Disorder Society, 29(13), 1606-1614 (2014-01-01)
Parkinson's disease (PD) is a neurodegenerative disorder for which environmental factors influence disease risk and may act via an epigenetic mechanism. The microtubule-associated protein tau (MAPT) is a susceptibility gene for idiopathic PD. Methylation levels were determined by pyrosequencing of
Seung-Ju Yang et al.
European journal of pharmacology, 736, 26-34 (2014-05-07)
Thiazole derivatives are attractive candidates for drug development because they can be efficiently synthesized and are active against a number of diseases and conditions, including diabetes. In our present study, we investigated the anti-inflammatory and antioxidant properties of N-adamantyl-4-methylthiazol-2-amine (KHG26693)
Sam B Weber et al.
Proceedings. Biological sciences, 279(1731), 1077-1084 (2011-09-23)
The effect of climate warming on the reproductive success of ectothermic animals is currently a subject of major conservation concern. However, for many threatened species, we still know surprisingly little about the extent of naturally occurring adaptive variation in heat-tolerance.
Siddharth Singh et al.
Clinical gastroenterology and hepatology : the official clinical practice journal of the American Gastroenterological Association, 13(4), 643-654 (2014-04-29)
Little is known about differences in rates of fibrosis progression between patients with nonalcoholic fatty liver (NAFL) vs nonalcoholic steatohepatitis (NASH). We conducted a systematic review and meta-analysis of all studies that assessed paired liver biopsy specimens to estimate the
Sarwat Jahan et al.
Systems biology in reproductive medicine, 60(4), 199-205 (2014-04-25)
The present study was performed to determine the effects of different antioxidants on testicular histopathology and oxidative damage induced by cadmium (Cd) in rat testis and prostate. Twenty five rats were equally divided into five groups (n = 5/group). The control group

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service