Skip to Content
Merck
All Photos(1)

Key Documents

85325

Sigma-Aldrich

Silver trifluoromethanesulfonate

purum, ≥98.0% (Ag)

Synonym(s):

Ag(OTf), Silver (trifluoromethyl)sulfonate, Silver triflate, Trifluoromethanesulfonic acid silver salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3SO3Ag
CAS Number:
Molecular Weight:
256.94
Beilstein:
3598402
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

grade

purum

Quality Level

Assay

≥98.0% (Ag)

form

crystals

reaction suitability

core: silver
reagent type: catalyst

SMILES string

[Ag+].[O-]S(=O)(=O)C(F)(F)F

InChI

1S/CHF3O3S.Ag/c2-1(3,4)8(5,6)7;/h(H,5,6,7);/q;+1/p-1

InChI key

QRUBYZBWAOOHSV-UHFFFAOYSA-M

General description

Silver trifluoromethanesulfonate p-complexes of monoenes, dienes, trienes, monoynes and diynes have been prepared. It reacts with 2-fluoro- and 3-fluoro-4-alkoxystilbazoles to afford the mesomorphic complexes. Iodine monochloride/AgOTf constitutes an efficient promoter system for the O-glycoside synthesis.

Application

Silver trifluoromethanesulfonate (AgOTf) may be employed as a reagent during glucosylation of several alcohols. AgOTf in combination with p-nitrobenzenesulfenyl chloride may be employed as an activator for the glycosylation.
It may be used for the synthesis of the following:
  • cystine-containing peptides
  • 3-aminoalkylated indoles
  • benzo[b]oxepines and 2H-chromenes
  • diversely substituted iminoimidazoazines

Other Notes

Reagent for the substitution of halides by triflate.; Reagent for the glycosylation of glycosyl halides; Reagent used for the deprotection of protected thiols

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Disulfide bond formation in S-acetamidomethyl cysteine-containing peptides by the combination of silver trifluoromethanesulfonate and dimethylsulfoxide/aqueous HCl.
Tamamura H, et al.
Tetrahedron Letters, 34(31), 4931-4934 (1993)
Silver Triflate Catalyzed Cyclopropyl Carbinol Rearrangement for Benzo [b] oxepine and 2H-Chromene Synthesis.
Chan PWH, et al.
European Journal of Organic Chemistry, 2015(20), 4447-4456 (2015)
The glucosylation of several alcohols with tetra-O-benzyl-. ALPHA.-D-glucopyranose and a mixture of p-nitrobenzenesulfonyl chloride, silver trifluoromethanesulfonate, and triethylamine.
Koto S, et al.
Bulletin of the Chemical Society of Japan, 53(6), 1761-1762 (1980)
V. Pozsgay et al.
The Journal of Organic Chemistry, 46, 3761-3761 (1981)
David Crich et al.
Carbohydrate research, 343(10-11), 1858-1862 (2008-04-01)
p-Nitrobenzenesulfenyl chloride is a stable commercially available sulfenyl chloride that, in conjunction with silver triflate, cleanly activates a wide range of thioglycosides for glycosylation at -78 degrees C in CH(2)Cl(2).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service