Skip to Content
Merck
All Photos(2)

Key Documents

84100

Millipore

Sucrose

ACS reagent, suitable for microbiology, ≥99.0%

Synonym(s):

α-D-Glc-(1→2)-β-D-Fru, α-D-Glucopyranosyl β-D-fructofuranoside, β-D-Fructofuranosyl-α-D-glucopyranoside, D(+)-Saccharose, Sugar

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H22O11
CAS Number:
Molecular Weight:
342.30
Beilstein:
90825
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

grade

ACS reagent

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D 66.3 to 66.8°, c = 26% in H2O

shelf life

limited shelf life, expiry date on the label

packaging

pkg of 1 kg
pkg of 250 g
pkg of 5 kg

storage condition

(Tightly closed)

impurities

≤0.0008 meq/g free acid
≤0.005% water insoluble matter
≤0.05% invert sugar
≤5 mg/kg heavy metals (as Pb)

ign. residue

≤0.01% (as SO4)

loss

≤0.03% loss on drying, 105 °C

pH

7.0 (20 °C, 100 g/L in water)

mp

185-187 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg
sulfate, sulfite (as SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

SMILES string

OC[C@H]1O[C@H](O[C@]2(CO)O[C@H](CO)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C12H22O11/c13-1-4-6(16)8(18)9(19)11(21-4)23-12(3-15)10(20)7(17)5(2-14)22-12/h4-11,13-20H,1-3H2/t4-,5-,6-,7-,8+,9-,10+,11-,12+/m1/s1

InChI key

CZMRCDWAGMRECN-UGDNZRGBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Sucrose can be used as a carbon source in microbial growth media. It provides a source of energy and carbon for bacteria and supports their growth. It is used to prepare several types of media for the isolation and cultivation of various bacteria, fungi, and yeasts. Sucrose is commonly used in media for the identification and differentiation of various bacteria that grow on sucrose-containing media. Chromogenic agar media, such as MacConkey agar media with sucrose, are also commonly used in the identification of bacterial strains. Sucrose can also be used in specialized media for the isolation and culture of certain organisms, such as lactic acid bacteria, which are commonly used in the food industry. Additionally, sucrose can serve as a differential reagent in media formulations, such as TSI test (triple sugar iron test), which differentiates various Enterobacteriaceae bacteria based on their ability to ferment various types of sugars.

Application

Sucrose is commonly used as an ingredient in many microbial culture media for the growth and identification of bacteria, yeasts, and molds from various pharmaceutical, environmental, and food and beverage samples.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Can sonication enhance release from liquid-core capsules
3 with a hydrogel membrane?
Liguo Zhang
Journal of Colloid and Interface Science (2011)
D. H. Lewis
Storage Carbohydrates in Vascular Plants: Distribution, Physiology and Metabolism, 53 -53 (1984)
Quantitative 1H NMR with external standards: use in preparation of calibration solutions for algal toxins and other natural products.
Burton IW, Quilliam MA, Walter JA.
Analytical Chemistry, 77(10), 3123-3131 (2005)
Denise Harrison et al.
Pediatrics, 130(5), 918-925 (2012-10-10)
The use of oral sucrose has been the most extensively studied pain intervention in newborn care to date. More than 150 published studies relating to sweet-taste-induced calming and analgesia in human infants have been identified, of which 100 (65%) include
Rafaela Ribeiro Reis et al.
Plant science : an international journal of experimental plant biology, 221-222, 59-68 (2014-03-25)
Drought is one of the most challenging agricultural issues limiting sustainable sugarcane production and, in some cases, yield losses caused by drought are nearly 50%. DREB proteins play vital regulatory roles in abiotic stress responses in plants. The transcription factor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service