Skip to Content
Merck
All Photos(2)

Documents

83400

Millipore

D-(+)-Raffinose pentahydrate

suitable for microbiology, ≥99.0%, Microbiological media component

Synonym(s):

O-α-D-Galactopyranosyl-(1-6)-α-D-glucopyranosyl β-D-fructofuranoside pentahydrate, Melitose, Melitriose, O-α-D-Galactopyranosyl-(1→6)-α-D-glucopyranosyl β-D-fructofuranoside, Melitose, Melitriose

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H32O16 · 5H2O
CAS Number:
Molecular Weight:
594.51
Beilstein:
99541
EC Number:
MDL number:
UNSPSC Code:
41106212
PubChem Substance ID:
NACRES:
NA.85

Quality Level

Assay

≥99.0% (HPLC)
≥99.0%

form

powder

optical activity

[α]20/D +105±2°, c = 10% in H2O

ign. residue

≤0.1% (as SO4)

mp

78-80 °C

solubility

H2O: 0.1 g/mL, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

As: ≤0.1 mg/kg
Ca: ≤20 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

microbiology

storage temp.

10-30°C

SMILES string

O.O.O.O.O.OC[C@H]1O[C@H](OC[C@H]2O[C@H](O[C@]3(CO)O[C@H](CO)[C@@H](O)[C@@H]3O)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C18H32O16.5H2O/c19-1-5-8(22)11(25)13(27)16(31-5)30-3-7-9(23)12(26)14(28)17(32-7)34-18(4-21)15(29)10(24)6(2-20)33-18;;;;;/h5-17,19-29H,1-4H2;5*1H2/t5-,6-,7-,8+,9-,10-,11+,12+,13-,14-,15+,16+,17-,18+;;;;;/m1...../s1

InChI key

BITMAWRCWSHCRW-PFQJHCPISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Raffinose is a natural trisaccharide composed of glucose, galactose, and fructose. It is a water-soluble oligosaccharide, most commonly present in legumes but also in sugar beets and remains during the process in the molasse. It is primarily used as a carbon source in the cultivation of yeast but also has certain cell proliferative and anti-oxidative properties that can prove useful in culture media. Raffinose is used by beneficial probiotic bacteria and an essential component of certain cryoprotectant solutions.

Application

D-(+)-Raffinose pentahydrate can be used as a carbon source for the cultivation of yeast
It is used by beneficial probiotic bacteria like Bifidobacteria and Lactobacilli but could be used as well for differentiation and identification of Escherichia coli, Enterococcus faecium and Streptococcus pneumoniae.
It can also be used in certain cryoprotectant solutions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Poonam Bheda et al.
STAR protocols, 1(3), 100228-100228 (2020-12-31)
The budding yeast Saccharomyces cerevisiae is an excellent model organism to dissect the maintenance and inheritance of phenotypes due to its asymmetric division. This requires following individual cells over time as they go through divisions to define pedigrees. Here, we
Poonam Bheda et al.
Molecular cell, 78(5), 915-925 (2020-05-12)
Transcriptional memory of gene expression enables adaptation to repeated stimuli across many organisms. However, the regulation and heritability of transcriptional memory in single cells and through divisions remains poorly understood. Here, we combined microfluidics with single-cell live imaging to monitor
Vidhya Bharathi et al.
Bio-protocol, 7(15), e2440-e2440 (2017-08-05)
The yeast Saccharomyces cerevisiae (S. cerevisiae) harboring ade1 or ade2 mutations manifest red colony color phenotype on rich yeast medium YPD. In these mutants, intermediate metabolites of adenine biosynthesis pathway are accumulated. Accumulated intermediates, in the presence of reduced glutathione
Antonio Marsella et al.
Cell reports, 34(13), 108906-108906 (2021-04-01)
The Mre11-Rad50-Xrs2 (MRX) complex detects and processes DNA double-strand breaks (DSBs). Its DNA binding and processing activities are regulated by transitions between an ATP-bound state and a post-hydrolysis cutting state that is nucleolytically active. Mre11 endonuclease activity is stimulated by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service