Skip to Content
Merck
All Photos(1)

Key Documents

83073

Supelco

(+)-α-Terpineol

analytical standard

Synonym(s):

(R)-2-(4-Methyl-3-cyclohexenyl)isopropanol, (R)-p-Menth-1-en-8-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2041428
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (sum of enantiomers, GC)

optical activity

[α]/D +90±10°, c = 0.1 in ethanol

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

density

0.94 g/mL at 20 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

CC1=CC[C@@H](CC1)C(C)(C)O

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3/t9-/m0/s1

InChI key

WUOACPNHFRMFPN-VIFPVBQESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: cinnamomum mentha pimpinella salvia thymus valeriana glycyrrhiza

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

194.0 °F - closed cup

Flash Point(C)

90 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gardênia Martins de Sousa et al.
Heliyon, 6(4), e03752-e03752 (2020-05-10)
Alpha-terpineol is a monoterpenoid found in many essential oils, being widely used in food and household products. In vitro antioxidant and anti-inflammatory activities have already been associated with this alcohol; therefore, this study aimed to check if these properties were
Mariusz Dziadas et al.
Acta scientiarum polonorum. Technologia alimentaria, 10(1), 7-17 (2012-01-11)
Amount of monoterpene alcohol and their glycoside precursors in grapes is determined by the type of grapes, and the availability of specific hydrolytic enzymes during the manufacturing process. Addition of these enzymes, hydrolysing β-glycosidic bond, to the grape can be
Soon-Nang Park et al.
Anaerobe, 18(3), 369-372 (2012-04-28)
Linalool and α-terpineol exhibited strong antimicrobial activity against periodontopathic and cariogenic bacteria. However, their concentration should be kept below 0.4 mg/ml if they are to be used as components of toothpaste or gargling solution. Moreover, other compounds with antimicrobial activity
Nasrin Samadi et al.
Natural product research, 26(20), 1931-1934 (2011-10-20)
The essential oil obtained from the flowering parts of Anthemis altissima L. var. altissima was analysed by gas chromatography and gas chromatography mass spectroscopy. In this study, 34 compounds representing 98.76% of the essential oil were identified. The main components
Chokri Messaoud et al.
Chemistry & biodiversity, 8(2), 300-310 (2011-02-22)
Extracts of mature dark blue and white berries from two Tunisian Myrtus communis morphs growing at the same site were assessed for their essential-oil and fatty-acid compositions, phenolic contents, and antioxidant activities. The GC and GC/MS analyses of the essential

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service