Recommended Products
description
Irreversible, single use
quality
Quality Assurance: Produced under ISO9001:2008
shelf life
limited shelf life, expiry date on the label
storage condition
room temperature
parameter
12 h run-out window ((calibrated at +22 °C)
size
40 mm × 19 mm
General description
Timestrip® indicators are simple, visual reminders for any time driven event. They are small, self-adhesive labels that can be attached to or integrated into a product or device. They monitor elapsed time and indicate when key time markers and a pre-defined elapsed time have been reached.
Visit our Timestrip Web Portal to learn more
Visit our Timestrip Web Portal to learn more
Application
Manual activation button on top. A colored line appears to show its armed. The color progresses along time markers in the viewing window.
Packaging
80956-10EA: 10 labels in sachet
80956-100EA: 100 labels in sachet
80956-500EA: 500 labels in sachet
80956-100EA: 100 labels in sachet
80956-500EA: 500 labels in sachet
Components
Non-toxic, polymeric assembly multilayer and aluminum-plastic laminate encapsulating colored oils
Specifications
- Time Accuracy: ±15% in time under isothermal conditions
- Usage: calibrated for an average temperature environment of 22°C/72°F
Legal Information
Timestrip is a registered trademark of Timestrip Limited
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service