Skip to Content
Merck
All Photos(3)

Key Documents

79610

Sigma-Aldrich

Phosphorus pentoxide

desiccant, with moisture indicator

Synonym(s):

Sicapent®, Phosphoric anhydride, Phosphorus(V) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
P2O5
CAS Number:
Molecular Weight:
141.94
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
grade:
for analytical purposes
form:
powder, crystals or granules

grade

for analytical purposes

Quality Level

vapor density

4.9 (vs air)

vapor pressure

1 mmHg ( 384 °C)
10 mmHg ( 238 °C)

form

powder, crystals or granules

quality

with moisture indicator

pH

1.5 (20 °C, 10 g/L)

mp

340 °C (lit.)

density

2.3 g/mL at 25 °C (lit.)

SMILES string

O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3

InChI

1S/O10P4/c1-11-5-12(2)8-13(3,6-11)10-14(4,7-11)9-12

InChI key

YWEUIGNSBFLMFL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus pentoxide (P2O5) is a phosphorous oxide. It participates in Pictet-Gams synthesis. Phosphorus pentoxide supported on alumina has been used for solvent-free and microwave-assisted synthesis of 1, 5-benzodiazepine derivatives.

Application

Phosphorus pentoxide may be used in the synthesis of dichlorine heptoxide (Cl2O7).

Legal Information

Sicapent is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Radek Pohl et al.
Organic & biomolecular chemistry, 13(16), 4693-4705 (2015-03-24)
Systematic structure-activity studies on a diverse family of nucleoside phosphonic acids has led to the development of potent antiviral drugs such as HPMPC (CidofovirTM), PMEA (AdefovirTM), and PMPA (TenofovirTM), which are used in the treatment of CMV-induced retinitis, hepatitis B
Jae-Hyung Ahn et al.
Microbial ecology, 68(2), 271-283 (2014-04-01)
The effects of water-saving irrigation on emissions of greenhouse gases and soil prokaryotic communities were investigated in an experimental rice field. The water layer was kept at 1-2 cm in the water-saving (WS) irrigation treatment and at 6 cm in
Ana García et al.
Acta biomaterialia, 7(3), 1265-1273 (2010-10-19)
Herein we report for the first time the synthesis of three-dimensional scaffolds in the binary system SiO2-P2O5 exhibiting different scales of porosity: (i) highly ordered mesopores with diameters of ca. 4 nm; (ii) macropores with diameters in the 30-80 μm
Eagleson M.
Concise Encyclopedia Chemistry, 215-215 (1994)
Alumina/phosphorus pentoxide (APP) as an efficient reagent for the synthesis of 1, 5-benzodiazepines under microwave irradiation.
Kaboudin B and Navaee K.
Heterocycles, 1443-1446 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service