Skip to Content
Merck
All Photos(5)

Key Documents

795445

Sigma-Aldrich

Calcium carbonate

anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CaCO3
CAS Number:
Molecular Weight:
100.09
Beilstein:
8008338
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
ACS reagent
anhydrous
form:
powder
solubility:
H2O: insoluble
Pricing and availability is not currently available.

grade

ACS reagent
anhydrous

Quality Level

product line

Redi-Dri

Assay

≥99%

form

powder

quality

free-flowing

impurities

≤0.002 meq/g water-soluble titr. base
≤0.005% oxidizers
≤0.01% NH4OH pptd.
≤0.01% insol. dil. HCl

pH

8

solubility

H2O: insoluble

density

2.93 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤0.001%
fluoride (F-): ≤0.0015%
sulfate (SO42-): ≤0.01%

cation traces

Ba: ≤0.005%
Fe: ≤0.003%
K: ≤0.01%
Mg: ≤0.02%
NH4+: ≤0.003%
Na: ≤0.1%
Sr: ≤0.1%
heavy metals: ≤0.001% (by ICP-OES)

SMILES string

[Ca++].[O-]C([O-])=O

InChI

1S/CH2O3.Ca/c2-1(3)4;/h(H2,2,3,4);/q;+2/p-2

InChI key

VTYYLEPIZMXCLO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Calcium carbonate is an inorganic compound that is commonly used as a solid base catalyst in some organic transformations. It is also used as an acid scavenger and stabilizing agent for acid-sensitive compounds. Additionally, CaCO3 is the most widely used additive in plastic and elastomeric applications due to its wide availability.

Application


  • Spectrophotometric Analysis: Chromotropic acid disodium salt dihydrate is used in spectrophotometric methods for detecting and quantifying metal ions. Studies have shown its effectiveness in forming stable complexes with metal ions such as thorium, which can then be analyzed spectrophotometrically. This application is essential for environmental and industrial monitoring of metal contaminants (Issa et al., International Journal of Pharmaceutical Sciences Review and Research, 2022).



Features and Benefits

  • Increased efficiency – Eliminates time and effort of loosening hardened powders     
  • Safety – Eliminates tools of force used to break up clumps
  • Ecology – Reduces waste as it does not stick to the packaging
  • Economy – Faster preparation and solvation increase productivity and reduce costs     
  • Assured Quality – Excellent, expert-tested quality with no anti-caking agents      
  • Flexible volumes – Available from research amounts to scale-up quantities

Legal Information

Redi-Dri is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the size of the calcium carbonate particles? What are the minerals and what are their contents in calcium carbonate?

    1 answer
    1. The particle size for this product is not determined. The trace analysis of this product is reported in the lot specific Certificate of Analysis. A sample or lot specific Certificate may be accessed at the link below:
      https://www.sigmaaldrich.com/product/sial/795445#product-documentation.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service