Skip to Content
Merck
All Photos(1)

Key Documents

78095

Supelco

Rutin trihydrate

analytical standard

Synonym(s):

Quercetin-3-rutinoside trihydrate, Vitamin P trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O16 · 3H2O
CAS Number:
Molecular Weight:
664.56
Beilstein:
75455
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥94.0% (HPLC)

optical activity

[α]20/D +9±2°, c = 0.5% in absolute ethanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

O.O.O.C[C@@H]1O[C@@H](OC[C@H]2O[C@@H](OC3=C(Oc4cc(O)cc(O)c4C3=O)c5ccc(O)c(O)c5)[C@H](O)[C@@H](O)[C@@H]2O)[C@H](O)[C@H](O)[C@H]1O

InChI

1S/C27H30O16.3H2O/c1-8-17(32)20(35)22(37)26(40-8)39-7-15-18(33)21(36)23(38)27(42-15)43-25-19(34)16-13(31)5-10(28)6-14(16)41-24(25)9-2-3-11(29)12(30)4-9;;;/h2-6,8,15,17-18,20-23,26-33,35-38H,7H2,1H3;3*1H2/t8-,15+,17-,18+,20+,21-,22+,23+,26+,27-;;;/m0.../s1

InChI key

NLLBWFFSGHKUSY-JPRRWYCFSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Rutin trihydrate is a bioflavonoid, which may play a key role in the functioning of capillaries.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: achillea crataegus echinacea eucalyptus ginkgo glycyrrhiza hedera humulus hypericum lycium pimpinella primula sambucus serenoa

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Using on-line solid phase extraction for simultaneous determination of ascorbic acid and rutin trihydrate by sequential injection analysis
Legnerova.Z, et al.
Analytica Chimica Acta, 497, 165-174 (2003)
Anete Boroduske et al.
Industrial crops and products, 165, 113438-113438 (2021-03-24)
Berries and flowers of Sambucus nigra L. tree are well known for their ability to mitigate symptoms of upper respiratory disorders related to reported antiviral properties. Industrial application and commercial cultivation of S. nigra is largely limited to a few
Nattapon Supkamonseni et al.
Indian journal of experimental biology, 52(10), 965-971 (2014-10-28)
In vitro study revealed that pancreatic lipase inhibitory activity of C. asiatica extract was significantly higher than rutin but lower than orlistat, an anti-obesity drug. alpha-Amylase inhibitory activities of C. asiatica extract and rutin were significantly lower than acarbose, an
Snezana Agatonovic-Kustrin et al.
Journal of chromatography. A, 1385, 103-110 (2015-02-11)
The present study describes a simple high performance thin layer chromatographic (HPTLC) method for the simultaneous quantification of apigenin, chamazulene, bisabolol and the use of DPPH free radical as a post-chromatographic derivatization agent to compare the free radical scavenging activities

Questions

  1. what is the solubility of rutin trihydrate in dmso?

    1 answer
    1. This product is not tested for solubility in DMSO. However various sources have reported a solubility of 25 - 30 mg/mL in DMSO. Please note that this information has not been validated.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service