76545
Hybriscan™ I kit
E. coli, suitable for agriculture, environmental, food and beverages, microbiology and in situ hybriziation
Synonym(s):
E. coli HybriScan™I, FastScan E. coli
About This Item
Recommended Products
product name
HybriScan™I E. coli, suitable for microbiology
technique(s)
microbe id | in situ hybriziation: suitable
Quality Level
application(s)
agriculture
environmental
food and beverages
microbiology
storage temp.
2-8°C
suitability
Escherichia coli
coliforms
Related Categories
General description
Features and Benefits
- Sensitivity (after pre enrichment): 110 cfu/L
- Sensitivity (cfu/assay): 5000
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Carc. 1B - Eye Dam. 1 - Met. Corr. 1 - Muta. 2 - Resp. Sens. 1 - Skin Corr. 1A - Skin Sens. 1 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service