Skip to Content
Merck
All Photos(1)

Key Documents

76357

Sigma-Aldrich

Patent blue VF sodium salt

for microscopy

Synonym(s):

Acid blue 1 sodium salt, Disulfin blue VN 150 sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31N2NaO6S2
CAS Number:
Molecular Weight:
566.66
Colour Index Number:
42045
Beilstein:
4108121
EC Number:
MDL number:
UNSPSC Code:
41116121
PubChem Substance ID:
NACRES:
NA.21

grade

for microscopy

Quality Level

form

powder

suitability

suitable for microscopy

SMILES string

[Na+].CCN(CC)c1ccc(cc1)\C(=C2/C=CC(\C=C2)=[N+](\CC)CC)c3ccc(cc3S([O-])(=O)=O)S([O-])(=O)=O

InChI

1S/C27H32N2O6S2.Na/c1-5-28(6-2)22-13-9-20(10-14-22)27(21-11-15-23(16-12-21)29(7-3)8-4)25-18-17-24(36(30,31)32)19-26(25)37(33,34)35;/h9-19H,5-8H2,1-4H3,(H-,30,31,32,33,34,35);/q;+1/p-1

InChI key

SJEYSFABYSGQBG-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Pavlista et al.
Ceska gynekologie, 77(3), 251-254 (2012-07-12)
Lymphatic mapping is a method to find and preserve upper extremity lymphatics during axillary surgery (axilla clearance and sentinel node biopsy) in breast cancer patients. This may reduce the incidence of lymphedema. We examined on anatomical model, if the lymphatic
O Brenet et al.
Acta anaesthesiologica Scandinavica, 57(1), 106-111 (2012-12-12)
An increasing number of immediate hypersensitivity reactions (HSR) have been reported after the use of Patent Blue V (PBV) for breast cancer surgery. This is the first study to publish prospective data with systematic allergological assessment. We conducted a multicentre
David Pavlista et al.
Annals of surgical oncology, 19(11), 3410-3415 (2012-04-25)
This anatomic study details the lymphatic drainage of the upper extremity (UE) and breast, as well as its course in the axilla and its relation to axillary reverse mapping. Two aspects important for breast cancer surgery were followed: connection between
The value of Q-switched Nd:YAG laser after patent blue-enhanced lymphaticovenular anastomosis.
Benoit Ayestaray et al.
Journal of plastic, reconstructive & aesthetic surgery : JPRAS, 66(8), 1157-1158 (2013-04-23)
Vernon K Sondak et al.
Annals of surgical oncology, 20(2), 680-688 (2012-10-12)
[(99m)Tc]Tilmanocept is a CD206 receptor-targeted radiopharmaceutical designed for sentinel lymph node (SLN) identification. Two nearly identical nonrandomized phase III trials compared [(99m)Tc]tilmanocept to vital blue dye. Patients received [(99m)Tc]tilmanocept and blue dye. SLNs identified intraoperatively as radioactive and/or blue were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service