Skip to Content
Merck
All Photos(1)

Documents

76176

Supelco

Methyl palmitoleate

analytical standard

Synonym(s):

Methyl cis-9-hexadecenoate, Palmitoleic acid methyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH=CH(CH2)7COOCH3
CAS Number:
Molecular Weight:
268.43
Beilstein:
1726111
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)
n20/D 1.451

bp

180-183 °C/1 mmHg (lit.)

mp

−0.5-0.5 °C (lit.)

density

0.875 g/mL at 25 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCC\C=C/CCCCCCCC(=O)OC

InChI

1S/C17H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17(18)19-2/h8-9H,3-7,10-16H2,1-2H3/b9-8-

InChI key

IZFGRAGOVZCUFB-HJWRWDBZSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methyl palmitoleate is a methyl ester of the monounsaturated fatty acid, palmitoleic acid.

Application

This analytical standard can also be used as follows:
  • Simultaneous determination of fatty acids in bovine colostrum samples by GC-FID after their derivatization to ester forms using an acidic catalyst boron trifluoride
  • Fatty acid analysis of biodiesel obtained from Datura stramonium Linn. by gas chromatography coupled with mass spectrometry (GC-MS)

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarin A, et al. et al.
Biodiesel: Production and Properties (2012)
P W Wertz et al.
Journal of dermatological science, 1(1), 33-37 (1990-01-01)
The goal of the present study was to determine whether topically applied fatty acid esters enter into epidermal lipid metabolism. Of special interest with respect to epidermal function were the linoleate-rich O-acylsphingolipids thought to be essential in maintenance of the
Eleftheria Diakogiannaki et al.
The Journal of endocrinology, 194(2), 283-291 (2007-07-21)
Long-chain saturated and monounsaturated fatty acids differ in their propensity to induce beta-cell death in vitro with palmitate (C16:0) being cytotoxic, whereas palmitoleate (C16:1n-7) is cytoprotective. We now show that this cytoprotective capacity extends to a poorly metabolised C16:1n-7 derivative
C Rentel et al.
Electrophoresis, 20(12), 2329-2336 (1999-09-28)
Miniaturized separation techniques such as capillary electrochromatography (CEC), pressurized capillary electrochromatography (pCEC) and capillary high performance liquid chromatography (CHPLC) have been coupled to a new detection technique: coordination ion spray mass spectrometry (CIS-MS). Electrospray ionization (ESI) has found widespread applications
Xian-Guo Zou et al.
Journal of agricultural and food chemistry, 62(43), 10594-10603 (2014-10-10)
In the present study, a human milk fat substitute (HMFS) enriched in medium-chain fatty acids (MCFAs) was synthesized through acidolysis reaction from Cinnamomum camphora seed oil (CCSO) with oleic acid in a solvent-free system. A commercial immobilized lipase, Lipozyme RM

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service