Skip to Content
Merck
All Photos(1)

Key Documents

72638

Supelco

(±)-Citronellal

analytical standard

Synonym(s):

(±)-3,7-Dimethyl-6-octenal

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=CHCH2CH2CH(CH3)CH2CHO
CAS Number:
Molecular Weight:
154.25
Beilstein:
1720789
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.451 (lit.)

bp

207 °C (lit.)

density

0.857 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

2-8°C

SMILES string

CC(CC\C=C(\C)C)CC=O

InChI

1S/C10H18O/c1-9(2)5-4-6-10(3)7-8-11/h5,8,10H,4,6-7H2,1-3H3

InChI key

NEHNMFOYXAPHSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

This compound is commonly found in plants of the genus: melissa

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yathsoeung Ho et al.
Pharmaceuticals (Basel, Switzerland), 13(12) (2020-12-24)
Triple negative breast cancer is one of the most aggressive breast cancer type with abilities of early metastasis and chemoresistance. The tropical plant Citrus hystrix DC. has been reported to promote many biological activities including anticancer. However, the effect of
Walter S Leal et al.
Proceedings of the National Academy of Sciences of the United States of America, 110(46), 18704-18709 (2013-10-30)
The southern house mosquito, Culex quinquefasciatus, has one of the most acute and eclectic olfactory systems of all mosquito species hitherto studied. Here, we used Illumina sequencing to identify olfactory genes expressed predominantly in antenna, mosquito's main olfactory organ. Less
Satish Chandra Philkhana et al.
Chemical communications (Cambridge, England), 49(32), 3342-3344 (2013-03-19)
Concise and protecting-group free synthesis of ent-palmyrolide A and (-)-cis-palmyrolide A were achieved starting from commercially available (S)-citronellal. The key fragment of palmyrolide A, "(5S,7S)-7-hydroxy-5,8,8-trimethylnonanamide", which makes up the most challenging part of the target molecule, was prepared in just
M S Melo et al.
Pharmaceutical biology, 48(4), 411-416 (2010-07-22)
Citronellal is a monoterpene reported to be a major component of the essential oils in various aromatic species of plants. The present study evaluated the central nervous system depressant and antinociceptive properties of citronellal through behavioral experimental models. Following intraperitoneal
Lucindo Quintans-Júnior et al.
Journal of medicinal food, 14(6), 630-639 (2011-04-13)
Citronellal (CT) is a monoterpenoid and the major constituent of the mixture of terpenoids that give the citronella oil its lemon scent. Citronella oil is widely used around the world for various purposes and is mainly obtained from plants of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service