Skip to Content
Merck
All Photos(1)

Key Documents

71995

Sigma-Aldrich

Sodium tartrate dibasic dihydrate

purum p.a., ≥98.0% (NT)

Synonym(s):

L-(+)-Tartaric acid disodium salt, Disodium tartrate dihydrate, Sodium tartrate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Na2O6 · 2H2O
CAS Number:
Molecular Weight:
230.08
Beilstein:
6121732
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Assay:
≥98.0% (NT)
form:
solid

grade

purum p.a.

Quality Level

Assay

≥98.0% (NT)

form

solid

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤100 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤200 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

SMILES string

[Na+].[Na+].[H]O[H].[H]O[H].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.2Na.2H2O/c5-1(3(7)8)2(6)4(9)10;;;;/h1-2,5-6H,(H,7,8)(H,9,10);;;2*1H2/q;2*+1;;/p-2/t1-,2-;;;;/m1..../s1

InChI key

FGJLAJMGHXGFDE-DGFHWNFOSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Sodium tartrate dibasic dihydrate (L-(+)-Tartaric acid disodium salt, Sodium-L-tartrate dibasic dihydrate) is the sodium salt of tartaric acid. Mixtures of dimethyl sulfoxide and 1% aqueous solution of sodium-L-tartrate dibasic dihydrate have been employed for the resolution of various quaternary ammonium surfactants.

Application

Sodium tartrate dibasic dihydrate (L-(+)-Tartaric acid disodium salt, Sodium-L-tartrate dibasic dihydrate) may be used for the growth of crystals of xylanase C (XynC) from Gram-positive bacterium Bacillus subtilis. It may be used to compose the reservoir solution for the growth of crystals of PcpR (a LysR-type transcription factor from Sphingobium chlorophenolicum L-1).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Resolution of a Five-Component Mixture of Quaternary Ammonium Surfactants on Silica Gel 60 F 254 High Performance Thin Layer Chromatographic Plates.
Bhawani SA, et al.
Journal of Surfactants and Detergents, 14(2), 301-305 (2011)
Franz J St John et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 65(Pt 5), 499-503 (2009-05-02)
The recent biochemical characterization of the xylanases of glycosyl hydrolase family 5 (GH 5) has identified a distinctive endo mode of action, hydrolyzing the beta-1,4 xylan chain at a specific site directed by the position of an alpha-1,2-linked glucuronate moiety.
Robert P Hayes et al.
International journal of molecular sciences, 15(11), 20736-20752 (2014-11-15)
PcpR is a LysR-type transcription factor from Sphingobium chlorophenolicum L-1 that is responsible for the activation of several genes involved in polychlorophenol degradation. PcpR responds to several polychlorophenols in vivo. Here, we report the crystal structures of the inducer-binding domain

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service