Skip to Content
Merck
All Photos(1)

Key Documents

71635

Sigma-Aldrich

Sodium citrate dibasic sesquihydrate

purum p.a., ≥99.0% (T)

Synonym(s):

Citric acid disodium salt, Disodium hydrogen citrate sesquihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC(COOH)(CH2COONa)2 · 1.5H2O
CAS Number:
Molecular Weight:
263.11
Beilstein:
3770069
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
form:
crystals
solubility:
water: soluble 263.1 g/L
Pricing and availability is not currently available.

grade

purum p.a.

Quality Level

Assay

≥99.0% (T)

form

crystals

pH

4.6-5.2 (25 °C, 50 mg/mL in H2O)

solubility

water: soluble 263.1 g/L

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤50 mg/kg
Cd: ≤50 mg/kg
Co: ≤50 mg/kg
Cu: ≤50 mg/kg
Fe: ≤50 mg/kg
K: ≤100 mg/kg
Ni: ≤50 mg/kg
Pb: ≤50 mg/kg
Zn: ≤50 mg/kg

functional group

carboxylic acid

SMILES string

O.O.O.[Na+].[Na+].[Na+].[Na+].OC(=O)CC(O)(CC([O-])=O)C([O-])=O.OC(=O)CC(O)(CC([O-])=O)C([O-])=O

InChI

1S/C6H8O7.2Na.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;;;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);;;1H2/q;2*+1;/p-2

InChI key

XNZQCYSOYHAYII-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium citrate dibasic sesquihydrate is a colorless crystalline sodium salt.

Application

Sodium citrate dibasic sesquihydrate may be employed in the following studies:
  • In the Supel Que Citrate (EN) tubes for the quantification of pesticides in blackcurrants by triple quadrupole GC-QqQ-MS/MS [Gas chromatography coupled to tandem mass spectrometry (GC-MS/MS) coupled with triple quadrupole (QqQ) analyzer] method.[1]
  • Dispersive solid phase extraction (dSPE) method, commonly referred as QuEChERS.2
  • Preparation of RNA precipitation solution.[2]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Edward M Turk et al.
The Journal of biological chemistry, 285(12), 8585-8594 (2010-01-13)
Yeast Suv3p is a member of the DEXH/D box family of RNA helicases and is a critical component of the mitochondrial degradosome, which also includes a 3' --> 5' exonuclease, Dss1p. Defects in the degradosome result in accumulation of aberrant
Stanisław Walorczyk
Talanta, 120, 106-113 (2014-01-29)
A triple quadrupole GC-QqQ-MS/MS method was optimized for multiresidue analysis of over 180 pesticides in blackcurrants. The samples were prepared by using a modified quick, easy, cheap, effective, rugged and safe (QuEChERS) analytical protocol. To reduce matrix co-extractives in the
Mudasir Bhat et al.
Environmental monitoring and assessment, 187(7), 398-398 (2015-06-04)
Dissipation patterns were studied following two applications of difenoconazole (score 25% EC) at 300 and 600 g ai ha(-1) as single and double dose respectively on Golden Delicious and Starkrimson cultivars of apple. Samples were collected at 0, 3, 6
H Abdallah et al.
Journal of chromatography. A, 1355, 61-72 (2014-06-25)
A new high performance liquid chromatography-high resolution mass spectrometry (HPLC-HRMS) method was developed for a simultaneous multi-residue analysis of 22 sulfonamides (SAs) and their metabolites in edible animal (pig, beef, sheep and chicken) tissues. Sample preparation was optimized on the
Matthew Edwards et al.
Journal of chromatography. A, 1402, 110-123 (2015-06-02)
Comprehensive two-dimensional gas chromatography (GC×GC) is recognised as a powerful tool for the separation of complex mixtures of volatile and semi-volatile compounds. In the analysis of challenging samples containing highly concentrated, active analytes or those with complicated matrices, it is

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service