Skip to Content
Merck
All Photos(1)

Key Documents

70149

Millipore

Nutrient Broth

No. 3, NutriSelect® Plus, suitable for microbiology

Synonym(s):

Standard - Nutrient Broth

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
41171607
NACRES:
NA.74

Quality Level

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

meat extract, 1 g/L
peptone, 5 g/L
sodium chloride, 5 g/L
yeast extract, 2 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

agriculture
clinical testing
environmental
food and beverages
life science and biopharma
water monitoring

microbiology

suitability

nonselective for Escherichia coli
nonselective for Pseudomonas spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Looking for similar products? Visit Product Comparison Guide

Application

An inexpensive liquid medium which serves as base for the culture and accumulation of microorganisms.

Preparation Note

Add 13 g to 1 litre of distilled water. Mix well and distribute into final containers. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Ersoz et al.
Cellular and molecular biology (Noisy-le-Grand, France), 63(7), 69-75 (2017-08-26)
The aim of this study was to investigate the anti-proliferative, apoptotic, cytotoxic, and anti-oxidant effects of extracts from the lichen Cladonia pocillumon human breast cancer cells (MCF-7), and to characterize the anti-microbial features.  MCF-7 cells were treated with methanolic C.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service