Skip to Content
Merck
All Photos(1)

Key Documents

63380

Sigma-Aldrich

Dimethyl malonate

purum, ≥96.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2(COOCH3)2
CAS Number:
Molecular Weight:
132.11
Beilstein:
774261
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Quality Level

grade

purum

Assay

≥96.0% (GC)

refractive index

n20/D 1.413 (lit.)
n20/D 1.413

bp

180-181 °C (lit.)

mp

−62 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)CC(=O)OC

InChI

1S/C5H8O4/c1-8-4(6)3-5(7)9-2/h3H2,1-2H3

InChI key

BEPAFCGSDWSTEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H Dongmei et al.
Preparative biochemistry & biotechnology, 30(3), 231-240 (2000-08-05)
5-Methoxytryptamine and L-tryptophan methyl ester were acylated with malonic acid, dimethyl malonate, or succinic anhydride to produce the corresponding N,N'-dicarbonyltryptamine derivatives. The analgesic activity was evaluated by the tail flick test. All of the compounds exhibited desirable analgesic potency. This
R Padmakumar et al.
Analytical biochemistry, 214(1), 318-320 (1993-10-01)
A rapid and high-yield synthesis of methylmalonyl coenzyme A is reported. The two-step procedure involves preparation of the thiophenyl ester of methylmalonic acid using dicyclohexylcarbodiimide as a condensing agent, followed by transesterification with coenzyme A, to yield methylmalonyl coenzyme A
Reiko Fujita et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 126(2), 109-116 (2006-02-08)
The carbon-carbon bond-forming reactions of 1-methyl-2-methylthiophenylpyridinium iodides with active methylene compounds (such as dimethyl malonate, malononitrile) using NaH as a base gave the 2-substituted methylene-1,2-dihydropyridine derivatives.
M Coker et al.
Journal of inherited metabolic disease, 19(6), 743-751 (1996-01-01)
Total plasma odd-numbered long-chain fatty acids were analysed in patients with methylmalonic acidaemia (vitamin B12-responsive and unresponsive), combined methylmalonic acidaemia/homocystinuria (CblC), propionic acidaemia (both neonatal-onset and late-onset), biotinidase deficiency and holocarboxylase synthase deficiency, as well as in hospital controls. Total
K Hiroi et al.
Chemical & pharmaceutical bulletin, 48(3), 405-409 (2000-03-22)
The Lewis acid-mediated reactions of allene-ene compounds, derived from 3-methylcitronellal or dimethyl malonate, were carried out using various Lewis acids such as ethylaluminum dichloride, diethylaluminum chloride, titanium chloride, zinc chloride etherate, or boron trifluoride etherate, affording unexpectedly intramolecular [2+2]cycloaddition products

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service