Skip to Content
Merck
All Photos(1)

Key Documents

63174

Sigma-Aldrich

4-Maleimidobutyric acid

≥98.0% (T)

Synonym(s):

γ-Maleimidobutyric acid, 4-Maleimidobutanoic acid, N-(3-Carboxypropyl)maleimide, N-Maleoyl-4-aminobutyric acid, N-Maleoyl-GABA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H9NO4
CAS Number:
Molecular Weight:
183.16
Beilstein:
1455876
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98.0% (T)

mp

95-98 °C

SMILES string

OC(=O)CCCN1C(=O)C=CC1=O

InChI

1S/C8H9NO4/c10-6-3-4-7(11)9(6)5-1-2-8(12)13/h3-4H,1-2,5H2,(H,12,13)

InChI key

NCPQROHLJFARLL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Gamma-maleimidobutyric acid may be used as a spacer in the construction of drug and other types of bioconjugates. 4-Maleimidobutyric acid is used with N-hydroxysuccinimide ester as a bifunctional cross-linking agent.
Modification reagent for thiol groups in proteins

Other Notes

SH-label for the modification of peptides and proteins; Used for the preparation of a new bleomycin analog for enzyme immunoassay (EIA); probe for membrane SH-groups

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N-polymethylenecarboxymaleimides -- a new class of probes for membrane sulphydryl groups.
D G Griffiths et al.
FEBS letters, 134(2), 261-263 (1981-11-16)
Biochemical Society Transactions, 11, 753-753 (1983)
K Fujiwara et al.
Cancer research, 41(10), 4121-4126 (1981-10-01)
An antibody directed toward pepleomycin, a new antitumor antibiotic related structurally to bleomycin, has been produced in rabbits by immunization with a pepleomycin-protein conjugate which was prepared by a novel procedure of coupling pepleomycin to mercaptosuccinylated bovine serum albumin using
Yuan Yuan et al.
Frontiers in bioscience : a journal and virtual library, 13, 3768-3774 (2008-05-30)
Development of sensitive and selective methods to detect proteins at trace levels is of great biological importance. Via derivatization with a bifunctional cross-linker 4-maleimidobutyric acid N-hydroxysuccinimide ester (GMBS) and an electrochemical marker 11-ferrocenyl-1-undecanethiol (Fc-SH), voltammetric determination of surface-confined proteins electrostatically
Marie Pribylova et al.
International journal of pharmaceutics, 415(1-2), 175-180 (2011-06-15)
A new targeted conjugates in which paclitaxel was used as a cytostatic compound and an analog of the gonadotropin-releasing hormone (GnRH) as a targeting moiety were synthesized. The molecule of the peptide hormone GnRH was modified to allow its connection

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service