Skip to Content
Merck
All Photos(2)

Key Documents

62571

Supelco

Lithium ionophore VIII

Selectophore, function tested

Synonym(s):

N,N,N′,N′,N″,N″-Hexacyclohexyl-4,4′,4″-propylidynetris(3-oxabutyramide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C48H83N3O6
CAS Number:
Molecular Weight:
798.19
Beilstein:
9248138
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

100
200

product line

Selectophore

quality

function tested

SMILES string

CCC(COCC(=O)N(C1CCCCC1)C2CCCCC2)(COCC(=O)N(C3CCCCC3)C4CCCCC4)COCC(=O)N(C5CCCCC5)C6CCCCC6

InChI

1S/C48H83N3O6/c1-2-48(36-55-33-45(52)49(39-21-9-3-10-22-39)40-23-11-4-12-24-40,37-56-34-46(53)50(41-25-13-5-14-26-41)42-27-15-6-16-28-42)38-57-35-47(54)51(43-29-17-7-18-30-43)44-31-19-8-20-32-44/h39-44H,2-38H2,1H3

InChI key

QSTVDAZAPLNMPD-UHFFFAOYSA-N

General description

Visit our Sensor Applications portal to learn more.

Application

Selected application examples

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Lipophilic, neutral ionophore for lithium ion-selective solvent polymeric membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Performance of a highly lipophilic Li+ lonophore in solvent polymeric membranes
Bochenska, M., Simon, W.
Mikrochimica Acta, 102, 277-281 (1990)
Novel flow-through bulk optode for spectrophotometric determination of lithium in pharmaceuticals and saliva
Albero, M.I., et al.
Sensors and Actuators B, Chemical, 145(1), 133-138 (2010)
Konstantin M Mikhelson et al.
Analytical chemistry, 74(3), 518-527 (2002-02-13)
Lithium-selective electrodes with solvent polymeric membranes based on two different dicyclohexylamide neutral ionophores are studied systematically. The selectivity of lithium response is studied by means of the ordinary potentiometric experiments. Stability constants of lithium, sodium, and potassium ions with the
Preparation of di- and triamides and their application in ion-selective electrodes
Bochenska, M., et al.
J. Incl. Phenom. Mol. Recog. Chem., 10, 19-27 (1991)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service