Skip to Content
Merck
All Photos(1)

Documents

55264

Supelco

Ibuprofen-d3

analytical standard

Synonym(s):

α-Methyl-d3-4-(isobutyl)phenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13D3H15O2
CAS Number:
Molecular Weight:
209.30
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

isotopic purity

95 atom % D

Assay

≥98.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental
forensics and toxicology
pharmaceutical (small molecule)

format

neat

mass shift

M+3

SMILES string

[2H]C([2H])([2H])C(C(O)=O)c1ccc(CC(C)C)cc1

InChI

1S/C13H18O2/c1-9(2)8-11-4-6-12(7-5-11)10(3)13(14)15/h4-7,9-10H,8H2,1-3H3,(H,14,15)/i3D3

InChI key

HEFNNWSXXWATRW-HPRDVNIFSA-N

General description

Ibuprofen-d3 is an isotopically labeled compound of ibuprofen, belonging to the class of non-steroidal anti-inflammatory drugs (NSAIDs).

Application

Ibuprofen-d3 has been used as internal standard in the analysis of pharmaceutical residues in river water using high performance liquid chromatography coupled with mass spectrometry (HPLC/MS-MS).
Ibuprofen-d3 has been used as an internal standard in the analysis of pharmaceutical residues in river water using high performance liquid chromatography coupled with mass spectrometry (HPLC/MS-MS). Ibuprofen-d3 may be used as an internal standard for the determination of diclofenac (DCF) in environmental samples using chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Athanasios Mantas et al.
Pharmaceutics, 11(2) (2019-02-10)
The formulation of arylpropionic acid derivatives (profens), which are poorly soluble Biopharmaceutical Classification System (BCS) Type II drugs, has a strong impact on their therapeutic action. This article shows that heat-treated powder mixtures of free acid profens with high surface
Carolina Aristizabal-Ciro et al.
Environmental science and pollution research international, 24(8), 7335-7347 (2017-01-21)
In this work, the presence of selected emerging contaminants has been investigated in two reservoirs, La Fe (LF) and Rio Grande (RG), which supply water to two drinking water treatment plants (DWTPs) of Medellin, one of the most populated cities
Formation of diclofenac and sulfamethoxazole reversible transformation products in aquifer material under denitrifying conditions: batch experiments
Barbieri M, et al.
The Science of the Total Environment, 426, 256-263 (2012)
Jaime Nivala et al.
The Science of the total environment, 649, 1144-1156 (2018-10-13)
Six pilot-scale treatment wetlands treating municipal wastewater were monitored for classical wastewater parameters and selected Emerging Organic Compounds (EOCs): caffeine (CAF), ibuprofen (IBU), naproxen (NPX), benzotriazole (BTZ), diclofenac (DCL), acesulfame (ACE) and carbamazepine (CBZ) on a weekly basis over the
Hiroyuki Mano et al.
Environmental monitoring and assessment, 189(9), 442-442 (2017-08-10)
The objective of this work was to evaluate the biological effect of the secondary effluent (SE) of a wastewater treatment plant and reclaimed water treated via ultrafiltration (UF) followed by either reverse osmosis (RO) membrane filtration or nanofiltration (NF) to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service