Skip to Content
Merck
All Photos(1)

Documents

51633

Supelco

Methyl heptadecanoate

analytical standard

Synonym(s):

Heptadecanoic acid methyl ester, Methyl margarate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15COOCH3
CAS Number:
Molecular Weight:
284.48
Beilstein:
1782681
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

152-153 °C/0.05 mmHg (lit.)

mp

29.8-30.3 °C (lit.)

format

neat

functional group

ester

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCCCC(=O)OC

InChI

1S/C18H36O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18(19)20-2/h3-17H2,1-2H3

InChI key

HUEBIMLTDXKIPR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Simona Tarricone et al.
Lipids in health and disease, 19(1), 119-119 (2020-06-04)
The present study aimed to evaluate the nutritional proximate composition, some qualitative traits and fatty acid profile of meat from wild thrush, woodcock and starling hunted in Southern Italy in 2017 and 2018. Nutritive composition and physical traits of meat
Rosa Palagano et al.
Foods (Basel, Switzerland), 9(7) (2020-07-18)
The content of fatty acid ethyl esters (FAEEs) is one of the quality parameters to define if an olive oil can be classified as extra virgin as these compounds are considered markers for virgin olive oils obtained from poor-quality olives.
Yingyang Zhang et al.
Journal of food science, 79(9), E1695-E1703 (2014-09-13)
The thermal properties of cured meat are important for determining storage life and nutritional quality. However, few studies have focused on the thermal properties of dry-cured pork, particularly in relation to salt level and type. In order to study the
Yuki Kitahara et al.
Bioscience, biotechnology, and biochemistry, 78(2), 336-342 (2014-07-19)
Rhodosporidium toruloides is a lipid-producing yeast, the growth of which is severely suppressed when hydrolysates of lignocellulosic biomass are used as carbon source. This is probably due to the toxic substances, such as organic acids, furans, and phenolic compounds produced
Faxiang Xu et al.
STAR protocols, 2(3), 100740-100740 (2021-09-02)
Metabolic homeostasis is critical for cell pluripotency and differentiation in human embryonic stem cells (hESCs). It has been reported that metabolic changes specifically regulate cellular signaling during hESC differentiation. This protocol describes procedures for both cell culture and detection of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service