Skip to Content
Merck
All Photos(1)

Key Documents

50553

Sigma-Aldrich

α-Tri-Calcium phosphate

Reagent for transient & stable DNA transfections

Synonym(s):

α-TCP, α-Tricalcium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca3(PO4)2
CAS Number:
Molecular Weight:
310.18
EC Number:
MDL number:
UNSPSC Code:
41106502
PubChem Substance ID:
NACRES:
NA.52

grade

for molecular biology

Quality Level

form

powder

technique(s)

transfection: suitable

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

storage temp.

room temp

SMILES string

[Ca++].[Ca++].[Ca++].[O-]P([O-])([O-])=O.[O-]P([O-])([O-])=O

InChI

1S/3Ca.2H3O4P/c;;;2*1-5(2,3)4/h;;;2*(H3,1,2,3,4)/q3*+2;;/p-6

InChI key

QORWJWZARLRLPR-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

General description

α-tri-Calcium is a tasteless powder containing excess of calcium oxide. It is commonly used to introduce DNA and transfect eukaryotic cells for molecular biology experiments.
α-tri-Calcium phosphate (α-TCP) can be made by heating the low-temperature polymorph β-TCP. It can also be made by thermal crystallization of amorphous precursors with the appropriate composition above the transformation temperature. α-TCP is biocompatible as β-TCP. It is highly soluble and hydrolyses quickly to calcium-deficient hydroxyapatite. This makes α-TCP a beneficial constituent for formulating self-setting osteotransductive bone cements and biodegradable bioceramics and composites for bone repairing.

Application

For analysis of bone cement; as standard.
α-tri-Calcium phosphate has been used to transfect 293T and 231 cells with the PiggyBac LSS-mOrange- aspartate-glutamate-valine-aspartate (DEVD)-mKate2 transposon vector and in reporter verification studies.
Suitable for
  • transient and stable transfection of wide range of cell types
  • drug delivery systems

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Caspase-3 Reporter for Fluorescence Lifetime Imaging of Single-Cell Apoptosis
Buschhaus J, et al.
Cells, 7(6), 57-57 (2018)
alpha-Tricalcium phosphate: Synthesis, properties and biomedical applications
Carrodeguas R G and De A S
Acta Biomaterialia, 7(10), 3536-3546 (2011)
Timo Brandenburger et al.
Shock (Augusta, Ga.), 42(3), 234-238 (2014-07-01)
Remote ischemic preconditioning (RIPC) is an easily applicable method for protecting the heart against a subsequent ischemia and reperfusion (I/R) injury. However, the exact molecular mechanisms underlying RIPC are unknown. We examined the involvement of microRNAs (miRNAs) and in particular
S I Roohani-Esfahani et al.
Acta biomaterialia, 8(11), 4162-4172 (2012-07-31)
This is the first reported study to prepare highly porous baghdadite (Ca₃ZrSi₂O₉) scaffolds with and without surface modification and investigate their ability to repair critical-sized bone defects in a rabbit radius under normal load. The modification was carried out to
A Butscher et al.
Acta biomaterialia, 9(2), 5369-5378 (2012-10-17)
Powder based three-dimensional printing (3DP) allows great versatility in material and geometry. These characteristics make 3DP an interesting method for the production of tissue engineering scaffolds. However, 3DP has major limitations, such as limited resolution and accuracy, hence preventing the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service