Skip to Content
Merck
All Photos(1)

Key Documents

46046

Supelco

α-tert-Butyl-β-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol

PESTANAL®, analytical standard

Synonym(s):

Paclobutrazol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H20ClN3O
Molecular Weight:
293.79
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

mixture of stereo isomers

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture

format

neat

SMILES string

OC(C(C)(C)C)C(N1N=CN=C1)CC2=CC=C(Cl)C=C2

InChI

1S/C15H20ClN3O/c1-15(2,3)14(20)13(19-10-17-9-18-19)8-11-4-6-12(16)7-5-11/h4-7,9-10,13-14,20H,8H2,1-3H3

InChI key

RMOGWMIKYWRTKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-tert-Butyl-β-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol is a known plant growth regulator which is utilized to inhibit the alternate bearing habit of mango (Mangifera indica L.).

Application

α-tert-Butyl-β-(4-chlorobenzyl)-1H-1,2,4-triazole-1-ethanol may be used as a reference standard for the determination of the analyte in:
  • Apple juice samples by matrix solid-phase dispersion (MSPD) and gas chromatography-mass selective detection (GC-MSD) with electron impact (EI) ionization.
  • Processed fruits and vegetables by miniaturized extraction-partition and liquid chromatography-electrospray tandem mass spectrometry (LC-ESI-MS/MS) equipped with multiple reaction monitoring (MRM) detection.
  • Crops and soya grains by LC-ESI-MS/MS with MRM detection.

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Flam. Sol. 1 - Repr. 2

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther Carrera et al.
Plant physiology, 160(3), 1581-1596 (2012-09-04)
procera (pro) is a tall tomato (Solanum lycopersicum) mutant carrying a point mutation in the GRAS region of the gene encoding SlDELLA, a repressor in the gibberellin (GA) signaling pathway. Consistent with the SlDELLA loss of function, pro plants display
Xiaorong Sun et al.
International journal of molecular sciences, 20(9) (2019-05-08)
Internode length is an important agronomic trait affecting plant architecture and crop yield. However, few genes for internode elongation have been identified in tomato. In this study, we characterized an elongated internode inbred line P502, which is a natural mutant
Uptake of soil applied paclobutrazol in mango (Mangifera indica L.) and its persistence in fruit and soil.
Sharma D and Awasthi MD
Chemosphere, 60(2), 164-169 (2005)
Meixiang Han et al.
Frontiers in plant science, 8, 1649-1649 (2017-10-12)
Petal appearance is an important horticultural trail that is generally used to evaluate the ornamental value of plants. However, knowledge of the molecular regulation of petal growth is mostly derived from analyses of
Comprehensive multi-residue method for the target analysis of pesticides in crops using liquid chromatography-tandem mass spectrometry.
Hiemstra M and de Kok A
Journal of Chromatography A, 1154(1-2), 3-25 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service