Skip to Content
Merck
All Photos(1)

Key Documents

46012

Supelco

Pentachloroaniline

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H2Cl5N
CAS Number:
Molecular Weight:
265.35
Beilstein:
2806732
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1c(Cl)c(Cl)c(Cl)c(Cl)c1Cl

InChI

1S/C6H2Cl5N/c7-1-2(8)4(10)6(12)5(11)3(1)9/h12H2

InChI key

KHCZSJXTDDHLGJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yan Wang et al.
Ecotoxicology and environmental safety, 182, 109463-109463 (2019-07-28)
To eliminate pentachloronitrobenzene (PCNB) residue in PCNB-contaminated environment, the degradation potential of Pseudomonas putida QTH3 to PCNB was evaluated in this study. Peudomonas putida QTH3 could grow well in mineral salt medium (MSM) containing PCNB as sole carbon and was
Didem Okutman Tas et al.
Environmental science & technology, 40(14), 4467-4472 (2006-08-15)
The microbial reductive dechlorination kinetics of pentachloroaniline (PCA) and less chlorinated anilines (CAs) were investigated with a mixed, fermentative/ methanogenic culture. Batch dechlorination assays were performed with all available CAs at an initial concentration of 3 microM, and an incubation
Zainab Z Ismail et al.
Biodegradation, 21(1), 43-57 (2009-06-27)
The reductive dechlorination of pentachloroaniline (PCA) was investigated in the absence and presence of sulfate in batch assays using a PCA-dechlorinating mixed anaerobic culture with methanol as the external electron donor at neutral pH and 22 degrees C. PCA at
Y Fushiwaki et al.
The Science of the total environment, 92, 55-67 (1990-03-01)
Environmental pollution by pentachloronitrobenzene (PCNB) was investigated at Tsumagoi, an intensive farming area, where a great amount of PCNB has been applied. High concentrations of PCNB were detected in river water near an area of cabbage cultivation. Further, pentachloroaniline (PCA)
Adriana Longoria et al.
Chemosphere, 72(3), 485-490 (2008-04-29)
Peroxidase transformations of widely distributed pollutants, tetra- and penta-chlorinated phenols and anilines, were studied using different peroxidases. Chloroperoxidase from Caldariomyces fumago was able to transform tetra- and penta-chlorinated phenols and anilines, while horseradish peroxidase, lignin peroxidase from Phanerochaete chrysosporium and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service