Skip to Content
Merck
All Photos(2)

Key Documents

46001

Supelco

Benoxacor

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11Cl2NO2
CAS Number:
Molecular Weight:
260.12
Beilstein:
4190275
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CC1COc2ccccc2N1C(=O)C(Cl)Cl

InChI

1S/C11H11Cl2NO2/c1-7-6-16-9-5-3-2-4-8(9)14(7)11(15)10(12)13/h2-5,7,10H,6H2,1H3

InChI key

PFJJMJDEVDLPNE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Benoxacor is a dichloroacetamide herbicide safener, used to protect corn against injury from metolachlor. Its role as a safener involves inducing the enzymatic mechanism of chloroacetanilide detoxification in plants.

Application

Benoxacor may be used as a reference standard in the determination of benoxacor in wheat and soil samples using gas chromatography (GC) and high performance liquid chromatography (HPLC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sihong Liu et al.
The Science of the total environment, 761, 143273-143273 (2020-11-17)
Benoxacor, a chiral herbicide safener for S-metolachlor, has been detected in streams. However, the potential risk this poses to aquatic ecosystems is not clear. This study used zebrafish (Danio rerio) embryos as a model to assess the enantioselective toxicity of
Daniele Del Buono et al.
Journal of agricultural and food chemistry, 53(11), 4326-4330 (2005-05-26)
Butachlor is a chloroacetanilide herbicide successfully employed in weeding some important crops, and benoxacor is a safening compound able to induce the enzymatic mechanism of chloroacetanilide detoxification in plants. A practical method for a simultaneous detection of butachlor and benoxacor
Purification and characterization of a glutathione S-transferase from benoxacor-treated maize (Zea mays).
Irzyk PG and Fuerst PE
Plant Physiology, 102(3), 803-810 (1993)
G Kocsy et al.
Plant physiology, 127(3), 1147-1156 (2001-11-14)
With the aim of analyzing their protective function against chilling-induced injury, the pools of glutathione and its precursors, cysteine (Cys) and gamma-glutamyl-Cys, were increased in the chilling-sensitive maize (Zea mays) inbred line Penjalinan using a combination of two herbicide safeners.
Luciano Scarponi et al.
Journal of agricultural and food chemistry, 53(7), 2483-2488 (2005-03-31)
The action of safener benoxacor on the detoxification of terbuthylazine (TBA) in Zea mays and Festuca arundinacea was ascertained by the investigation of the effects of benoxacor on the activity of glutathione-S-transferases (GSTs) in the shoots of the two plant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service