Skip to Content
Merck
All Photos(2)

Key Documents

457078

Sigma-Aldrich

1-Pyrenebutyric acid N-hydroxysuccinimide ester

95%

Synonym(s):

N-Hydroxysuccinimidyl Pyrenebutanoate; 2,5-Dioxopyrrolidin-1-yl 4-(pyren-1-yl)butanoate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H19NO4
CAS Number:
Molecular Weight:
385.41
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Quality Level

Assay

95%

form

powder, crystals or chunks

composition

Carbon: 70.7-78.9%

technique(s)

titration: suitable

color

faint beige

mp

132-136 °C (lit.)

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

O=C(CCCc1ccc2ccc3cccc4ccc1c2c34)ON5C(=O)CCC5=O

InChI

1S/C24H19NO4/c26-20-13-14-21(27)25(20)29-22(28)6-2-3-15-7-8-18-10-9-16-4-1-5-17-11-12-19(15)24(18)23(16)17/h1,4-5,7-12H,2-3,6,13-14H2

InChI key

YBNMDCCMCLUHBL-UHFFFAOYSA-N

General description

1-Pyrenebutyric acid N-hydroxysuccinimide ester (PANHS) is an aromatic chemical, which is also known as PBSE. It is a yellow solid, that is soluble in chloroform, N,N dimethyl formamide, dimethyl sulfoxide, and methanol.

Application

1-Pyrenebutyric acid N-hydroxysuccinimide ester is used in detecting nucleic acids, characterizing cyclodextrin based polyrotaxanes, and immobilizing proteins etc.

Other Notes

1-Pyrenebutyric acid N-hydroxysuccinimide ester has been used for NAD+ tethering on multi-walled carbon nanotubes (MWCNTs). It has been used in the preparation of laccase cathode.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pei-Ying Lo et al.
Materials science & engineering. C, Materials for biological applications, 109, 110593-110593 (2020-04-02)
Giving patients right dosage is an essential concept of precision medicine. Most of nanocarriers lack of flexible drug capacity and structural stability to be customized for specific treatment, resulting in low therapeutic efficacy and unexpected side effects. Thus, a growing
Ziran Wang et al.
Nanomaterials (Basel, Switzerland), 10(8) (2020-08-06)
A wearable and deformable graphene-based field-effect transistor biosensor is presented that uses aptamer-modified graphene as the conducting channel, which is capable of the sensitive, consistent and time-resolved detection of cytokines in human biofluids. Based on an ultrathin substrate, the biosensor
Biljana Mojsoska et al.
Sensors (Basel, Switzerland), 21(2) (2021-01-13)
The outbreak of the coronavirus disease (COVID-19) pandemic caused by the novel coronavirus (SARS-CoV-2) has been declared an international public health crisis. It is essential to develop diagnostic tests that can quickly identify infected individuals to limit the spread of
Masami Naya et al.
International journal of molecular sciences, 21(20) (2020-10-18)
Fluorescence microscopy (FM) has revealed vital molecular mechanisms of life. Mainly, molecules labeled by fluorescent probes are imaged. However, the diversity of labeling probes and their functions remain limited. We synthesized a pyrene-based fluorescent probe targeting SH groups, which are
Nguyen Thanh Tung et al.
Scientific reports, 7(1), 17881-17881 (2017-12-22)
Biosensors employing single-walled carbon nanotube field-effect transistors (SWCNT FETs) offer ultimate sensitivity. However, besides the sensitivity, a high selectivity is critically important to distinguish the true signal from interference signals in a non-controlled environment. This work presents the first demonstration

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service