Skip to Content
Merck
All Photos(1)

Key Documents

45517

Supelco

Furathiocarb

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H26N2O5S
CAS Number:
Molecular Weight:
382.47
Beilstein:
8396473
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCCCOC(=O)N(C)SN(C)C(=O)Oc1cccc2CC(C)(C)Oc12

InChI

1S/C18H26N2O5S/c1-6-7-11-23-16(21)19(4)26-20(5)17(22)24-14-10-8-9-13-12-18(2,3)25-15(13)14/h8-10H,6-7,11-12H2,1-5H3

InChI key

HAWJXYBZNNRMNO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kwang-Hyeon Liu et al.
Archives of toxicology, 77(5), 255-260 (2003-05-08)
In this study, the dermal penetration rate of carbofuran, carbosulfan, and furathiocarb has been measured with rat abdominal skin using the static diffusion cell. The technical grades of three compounds were applied at different doses on skin surface mounted in
Tomoki Fukuyama et al.
Toxicology, 261(3), 152-161 (2009-05-27)
Several types of pesticides, such as organophosphates, phenoxyacetic acid, and carbamate have a high risk of affecting human health, causing allergic rhinitis and bronchial asthma-like diseases. We used our long-term sensitization method and a local lymph node assay to examine
S K Lee et al.
Forensic science international, 101(1), 65-70 (1999-06-22)
Seven cases involving acute fatalities due to ingestion of furathiocarb, a carbamate insecticide, are presented. Furathiocarb was detected in the gastric contents using thin layer chromatography (TLC) and gas chromatography/mass spectrophotometry (GC/MS), and quantified in the blood using a gas
[Sensitivity of taiga mites to dilor, etafos, izophen, omait and deltanit].
Iu S Korotkov et al.
Meditsinskaia parazitologiia i parazitarnye bolezni, (1)(1), 37-40 (1987-01-01)
P Stehrer-Schmid et al.
Mutation research, 345(3-4), 111-125 (1995-12-01)
The carbamate insecticides benfuracarb, carbosulfan and furathiocarb were investigated in the mouse bone marrow micronucleus assay to establish whether they show cytogenetic activity in vivo. Two doses of each substance were administered intraperitoneally to NMRI mice. All of the three

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service