Skip to Content
Merck
All Photos(1)

Key Documents

45515

Supelco

Fuberidazole

PESTANAL®, analytical standard

Synonym(s):

2-(2-Furanyl)-1H-benzimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H8N2O
CAS Number:
Molecular Weight:
184.19
Beilstein:
153833
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

c1ccc2[nH]c(nc2c1)-c3ccco3

InChI

1S/C11H8N2O/c1-2-5-9-8(4-1)12-11(13-9)10-6-3-7-14-10/h1-7H,(H,12,13)

InChI key

UYJUZNLFJAWNEZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 Inhalation - Skin Sens. 1 - STOT RE 2

Target Organs

Heart

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Antonia Moral et al.
Analytica chimica acta, 650(2), 207-213 (2009-09-02)
A supramolecular solvent consisting of vesicles, made up of equimolecular amounts of decanoic acid (DeA) and tetrabutylammonium decanoate (Bu4NDe), dispersed in a continuous aqueous phase, is proposed for the extraction of benzimidazolic fungicides (BFs) from fruits and vegetables. Carbendazim (CB)
Gisela N Piccirilli et al.
The Analyst, 135(6), 1299-1308 (2010-04-17)
This paper presents a novel approach for the simultaneous determination of two widely used fungicides in a very interfering environment, combining the advantage of a spectrofluorimetric optosensor coupled to a flow-injection system and the selectivity of second-order chemometric algorithms. The
D Picón Zamora et al.
The Analyst, 125(6), 1167-1174 (2000-08-10)
The simultaneous determination of carbendazim, fuberidazole and thiabendazole was accomplished by cross-section (CS) fluorimetry in combination with multivariate calibration algorithms. The total luminescence information of the compounds was used to optimise the linear trajectories of the CS. A comparison between
A Garrido Frenich et al.
Analytical and bioanalytical chemistry, 375(7), 974-980 (2003-04-23)
The application of the generalised rank annihilation method (GRAM) and the trilinear decomposition (TLD) method to the resolution and quantitation of fluorescence excitation-emission matrices of a ternary mixture of pesticides, carbendazim, fuberidazole, and thiabendazole, with overlapped spectra is described. The
Adrian Frank et al.
Basic & clinical pharmacology & toxicology, 96(4), 325-330 (2005-03-10)
During metabolism of the fungicide 2-(2-furyl)benzimidazole (FB) in mammals, an optically active compound, metabolite A, was isolated. This compound, a principal metabolite in the biological degradation of FB, was isolated from the urine of horse and dog. When studied by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service