Skip to Content
Merck
All Photos(2)

Key Documents

45334

Supelco

Aziprotryne

PESTANAL®, analytical standard

Synonym(s):

2-Azido-4-isopropylamino-6-methylthio-1,3,5-triazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N7S
CAS Number:
Molecular Weight:
225.27
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CSc1nc(NC(C)C)nc(N=[N+]=[N-])n1

InChI

1S/C7H11N7S/c1-4(2)9-5-10-6(13-14-8)12-7(11-5)15-3/h4H,1-3H3,(H,9,10,11,12)

InChI key

AFIIBUOYKYSPKB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomasz Rejczak et al.
Food chemistry, 217, 225-233 (2016-09-25)
In this study, a quick, easy, cheap, effective, rugged and safe (QuEChERS) extraction technique was adapted to develop a simple sample treatment for multi-residue pesticide analysis in milk samples. The proposed method is based on liquid-liquid partitioning with acetonitrile followed
A Zapardiel et al.
Fresenius' journal of analytical chemistry, 367(5), 461-466 (2001-03-03)
The detection and determination of s-triazines, atrazine-desethyl and aziprotryne by cyclic voltammetry and an amperometric method using a metallic copper electrode and a glassy carbon electrode are described. The concentrations of atrazine-desethyl and aziprotryne in 0.1 M NaOH solutions were
R Gálvez et al.
Fresenius' journal of analytical chemistry, 367(5), 454-460 (2001-03-03)
The electroanalytical behavior of the reduction of the herbicides aziprotryne (2-azido-4-isopropylamino-6-methylthio-1,3,5-triazine) and desmetryne (4-isopropylamino-6-methylamino-2-methylthio-1,3,5-triazine) in oil-in-water emulsions is reported. This medium allows the differential pulse polarographic determination of these s-triazines directly from their sample extracts in an appropriate organic solvent.
[Gas-chromatographic analysis of methylthio-containing triazines in biological substrates].
G Shumann et al.
Gigiena i sanitariia, (12)(12), 81-82 (1990-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service