Skip to Content
Merck
All Photos(1)

Documents

43905

Supelco

Terpinolene

analytical standard

Synonym(s):

4-Isopropylidene-1-methylcyclohexene, p-Menth-1,4(8)-diene, p-Meth-1-en-8-yl-formate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16
CAS Number:
Molecular Weight:
136.23
Beilstein:
1851203
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

~4.7 (vs air)

vapor pressure

~0.5 mmHg ( 20 °C)

Assay

≥94% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

refractive index

n20/D 1.487-1.491
n20/D 1.489 (lit.)

bp

184-185 °C (lit.)

density

0.861 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

C\C(C)=C1/CCC(C)=CC1

InChI

1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4H,5-7H2,1-3H3

InChI key

MOYAFQVGZZPNRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 3 - Asp. Tox. 1 - Skin Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

141.8 °F - Seta closed cup

Flash Point(C)

61 °C - Seta closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service