Skip to Content
Merck
All Photos(1)

Documents

43720

Sigma-Aldrich

N,N′-Disuccinimidyl carbonate

≥95.0% (NMR), for peptide synthesis

Synonym(s):

N-Succinimidyl carbonate, DSC, Di(N-succinimidyl) carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H8N2O7
CAS Number:
Molecular Weight:
256.17
Beilstein:
1499137
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

product name

N,N′-Disuccinimidyl carbonate, purum, ≥95.0% (NMR)

grade

purum

Quality Level

Assay

≥95.0% (NMR)

form

powder

reaction suitability

reaction type: Carbonylations

impurities

~3% N-hydroxysuccinimide (NMR)

mp

190 °C (dec.) (lit.)

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

O=C1CCC(=O)N1OC(=O)ON2C(=O)CCC2=O

InChI

1S/C9H8N2O7/c12-5-1-2-6(13)10(5)17-9(16)18-11-7(14)3-4-8(11)15/h1-4H2

InChI key

PFYXSUNOLOJMDX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N′-Disuccinimidyl carbonate (DSC) can be used to synthesize:
  • Various carbamate derivatives from primary and sterically hindered secondary alcohols by alkoxycarbonylation.
  • Active carbonate resins from 4-hydroxymethylpolystyrene and 4-hydroxymethyl-3-nitrobenzamido resins via hydroxy functional groups.
  • Aza-glycinyl dipeptides, important intermediates for the preparation of various azapeptides.

It may be also used:
  • In the two-step preparation of 5-(6-(azidomethyl)nicotinamido)pentanoic acid, a copper-chelating picolyl azide derivative.
  • To activate the hydroxyl group of the hapten, γ-hydroxyphenylbutazone (HPBZ) so that HPBZ can effectively bind with human serum albumin(HSA)-immunogen to form a hapten-protein conjugate.

Other Notes

Convenient reagent for preparing N-succinimidyl esters of N-protected amino acids and other acids; activated carbonate, e.g. synthesis of ureas and carbamates; coupling of ligands to proteins (via lysines).

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT RE 2 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N. Nimura et al.
Analytical Chemistry, 58, 2372-2372 (1986)
Arun K Ghosh et al.
Tetrahedron letters, 33(20), 2781-2784 (1992-05-12)
An efficient and mild method for alkoxycarbonylation of amines is described, utilizing commercially available N,N'-disuccinimidyl carbonate.
A.K. Ghosh et al.
Tetrahedron Letters, 33, 2781-2781 (1992)
Active carbonate resins for solid-phase synthesis through the anchoring of a hydroxyl function. Synthesis of cyclic and alcohol peptides
Alsina J, et al.
Tetrahedron Letters, 38(5), 883-886 (1997)
Yesica Garcia-Ramos et al.
Journal of peptide science : an official publication of the European Peptide Society, 19(12), 725-729 (2013-11-10)
Aza-glycinyl dipeptides are useful building blocks for the synthesis of a diverse array of azapeptides. The construction of the aza-glycine residue is however challenging, because of the potential for side reactions, such as those leading to formation of oxadiazalone, hydantoin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service