Skip to Content
Merck
All Photos(1)

Key Documents

43538

Supelco

Tridodecylamine hydrochloride

Selectophore, ≥98.0% (AT)

Synonym(s):

Tridodecylammonium chloride, Trilaurylammonium chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H75N· HCl
CAS Number:
Molecular Weight:
558.45
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:

product line

Selectophore

Quality Level

Assay

≥98.0% (AT)

form

solid

SMILES string

Cl.CCCCCCCCCCCCN(CCCCCCCCCCCC)CCCCCCCCCCCC

InChI

1S/C36H75N.ClH/c1-4-7-10-13-16-19-22-25-28-31-34-37(35-32-29-26-23-20-17-14-11-8-5-2)36-33-30-27-24-21-18-15-12-9-6-3;/h4-36H2,1-3H3;1H

InChI key

GPQCSCQDQNXQSV-UHFFFAOYSA-N

General description

Tridodecylamine hydrochloride (TDA-HCl) is a hydrophobic tertiary amine.
Visit our Sensor Applications portal to learn more.

Application

It may be used be as buffer dissolved in toluene and hexane during transesterification of N-Ac-L-TyrOEt with PrOH catalyzing the activity of subtilisin Carlsberg.

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tridodecylamine and its hydrochloride salt: an organo-soluble buffer for subtilisin Carlsberg in non-polar organic solvents.
Harper, Neil, Barry D. Moore, and Peter J. Halling
Tetrahedron Letters, 41.21, 4223-4227 (2000)
J M Rawlings et al.
Gut, 26(2), 203-207 (1985-02-01)
Plastic electrodes have been developed for measuring pH in the human gastrointestinal tract. The electrodes have a plastic hydrogen ion sensitive membrane sealed to a length of fluid filled PVC tubing. Two recently developed hydrogen ion sensitive ligands have been
G T McEwan et al.
Journal of medical engineering & technology, 14(1), 16-20 (1990-01-01)
Incorporation of a ligand into polyvinylchloride allows the production of ion selective electrodes that are soft-bodied and disposable. With features make them especially suitable for clinical, particularly gastroenterological, investigations. We report here on the construction of a combined reference and
M O'Dowd et al.
Journal of biomedical engineering, 10(2), 165-170 (1988-04-01)
Frequent measurement of pH and PCO2 levels of fetal blood during labour provides an insight into the presence and nature of fetal asphyxia, effectiveness of in utero resuscitation and the need for operative delivery. A pH measuring system utilizing a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service