Skip to Content
Merck
All Photos(1)

Documents

433942

Supelco

Amberlite HPR4800 Ion Exchange Resin

chloride form, 25-35 mesh

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
SB.52

product name

AmberLite HPR4800 Cl, chloride form,

description

chloride form
formerly Dowex Marathon A, chloride form

Quality Level

form

beads

crosslinking

8‑10 % cross-linked

quality

monodispersed

moisture

50-60%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

quaternary ammonium

particle size

24-28 mesh
525-625 μm

capacity

1.3 meq/mL by wetted bed volume

separation technique

anion exchange

General description

Dowex Marathon A is a strong basic anionic Type I exchange gel resin with trimethylamine as functional group. Dowex Marathon A is a heterogeneous catalyst made of styerenedivinylbenzene copolymer.

Application

Dowex Marathon A was used in ion-exchange chromatography.
Strongly basic (type I) anion exchange resin for demineralization, especially suited for water with high concentration of weak ions (silica, CO2), as well as organics-laden feed waters. This resin meets requirements of FDA Food Additive Regulation 21 CFR 173.25.

Features and Benefits

Strongly basic anion

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.
Marathon is a trademark of The Dow Chemical Company or an affiliated company of Dow

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The synergistic effect of alcohol mixtures on transesterification of soybean oil using homogeneous and heterogeneous catalysts
Kim, M., et al.
Applied Catalysis A: General, 378, 134-143 (2010)
Dissolved organic matter removal and disinfection byproduct formation control using ion exchange
Tan, Yongrui, et al.
Desalination, 176, 189-200 (2005)
Synthetic route to a benzooxazole derivative with heparanase inhibitory activity
Da Pieve, Chiara, Pravin Patel, and Sotiris Missailidis
Synthetic Communications, 40, 518-522 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service