Skip to Content
Merck
All Photos(1)

Key Documents

42993

Supelco

Theobromine

analytical standard

Synonym(s):

2,6-Dihydroxy-3,7-dimethylpurine, 3,7-Dihydro-3,7-dimethyl-1H-purine-2,6-dione, 3,7-Dimethylxanthine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8N4O2
CAS Number:
Molecular Weight:
180.16
Beilstein:
16464
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

345-350 °C

suitability

conforms to structure for Infrared spectrum

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

[H]N1C(N(C)C2=C(N(C)C=N2)C1=O)=O

InChI

1S/C7H8N4O2/c1-10-3-8-5-4(10)6(12)9-7(13)11(5)2/h3H,1-2H3,(H,9,12,13)

InChI key

YAPQBXQYLJRXSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Theobromine is an active bronchodilator drug ingredient used in the treatment of acute and chronic asthma.[1] It is a naturally present alkaloid in a wide variety of foods and beverages, such as coffee, cocoa, tea, yerba mate and energizing drinks.[2]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Theobromine may be used as a reference standard for the determination of theobromine metabolites in human plasma and urine after consumption of soluble cocoa products with different methylxanthine contents by high-performance liquid chromatography and diode array detection (HPLC-DAD) method.[2]

Biochem/physiol Actions

Phosphosdiesterase inhibitor; weak adenosine receptor antagonist; diuretic; smooth muscle relaxant.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Theobromine, caffeine, and theophylline metabolites in human plasma and urine after consumption of soluble cocoa products with different methylxanthine contents.
Martinez-Lopez S.
Food Research International, 63, 446-455 (2014)
Rita Petrucci et al.
Biochimica et biophysica acta. General subjects, 1862(8), 1781-1789 (2018-05-16)
Antioxidant properties have been recently suggested for caffeine that seems showing protective effects against damages caused by oxidative stress. In particular, a HO scavenging activity has been ascribed to caffeine. Even if the oxidation of caffeine has been widely studied
Marcia S Bispo et al.
Journal of chromatographic science, 40(1), 45-48 (2002-03-02)
This work relates the development of an analytical methodology to simultaneously determine three methylxanthines (caffeine, theobromine, and theophylline) in beverages and urine samples based on reversed-phase high-performance liquid chromatography. Separation is made with a Bondesil C18 column using methanol-water-acetic acid
Anne-Laure Gassner et al.
Journal of chromatography. A, 1353, 121-130 (2014-06-14)
Procedures for estimating the measurement uncertainty (MU) of the concentration of a given analyte in a sample are of major concern for analytical chemists. Unfortunately, it is still unclear how and why MU should be assessed. While several possibilities exist
Takashi Koyanagi et al.
Xenobiotica; the fate of foreign compounds in biological systems, 45(4), 312-321 (2014-11-05)
1. Pharmacokinetics of human cytochrome P450 probes (caffeine, racemic warfarin, omeprazole, metoprolol and midazolam) were investigated after single intravenous and oral administrations at doses of 0.20 and 1.0 mg kg(-1), respectively, in combination to three young (3-year-old) and three aged (16-year-old) cynomolgus

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service