Skip to Content
Merck
All Photos(1)

Key Documents

40405

Millipore

Hippurate Disks

suitable for microbiology

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171621
NACRES:
NA.85

product line

BioChemika

shelf life

limited shelf life, expiry date on the label

technique(s)

microbe id | specific enzyme detection: suitable

application(s)

clinical testing
food and beverages

microbiology

storage temp.

2-8°C

suitability

Campylobacter spp.
Gardnerella vaginalis
Streptococcus spp.

General description

Recommended in qualitative procedures to detect organisms possessing the enzyme hippurate hydrolase, which promotes the hydolysis of the peptide bond in hippurate, releasing glycine and benzoic acid as end-products. The disk method is a rapid test useful for differentiating beta-hemolytic group B streptococci, Gardnerella vaginalis and Campylobacter species.

Packaging

Sterile filter paper discs (25 pcs, diameter 10 mm) impregnated with sodium hippurate

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service