Skip to Content
Merck
All Photos(1)

Key Documents

39068

Sigma-Aldrich

4-(Dimethylamino)azobenzene-4′-sulfonyl chloride

≥97.5% (AT)

Synonym(s):

4-(4-Dimethylaminophenylazo)benzenesulfonyl chloride, DABS-Cl, Dabsyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4N=NC6H4SO2Cl
CAS Number:
Molecular Weight:
323.80
Beilstein:
3064095
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥97.5% (AT)

mp

185 °C (dec.) (lit.)

solubility

DMF: soluble
acetonitrile: soluble

SMILES string

CN(C)c1ccc(cc1)\N=N\c2ccc(cc2)S(Cl)(=O)=O

InChI

1S/C14H14ClN3O2S/c1-18(2)13-7-3-11(4-8-13)16-17-12-5-9-14(10-6-12)21(15,19)20/h3-10H,1-2H3/b17-16+

InChI key

VTVWTPGLLAELLI-WUKNDPDISA-N

Looking for similar products? Visit Product Comparison Guide

General description

Dabsyl chloride (4-dimethylaminoazobenzene-4′-sulfonyl chloride) is a chromophoric labeling reagent used in HPLC for derivatizing amino acids. It reacts freely with all amino acids in order to form dabsyl amino acids, which are photostable and can be seen on a thin-layer chromatographic plate.

Application

For the qualitative and quantitative identification of amino acids and for the determination of N-terminal amino acids. Used in the determination of primary and secondary amines by HPLC.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Chromophoric labeling of amino acids with 4-dimethylaminoazobenzene-4'-sulfonyl chloride.
J K Lin et al.
Analytical chemistry, 47(9), 1634-1638 (1975-08-01)
S E Plyte et al.
Biochemistry, 32(14), 3623-3628 (1993-04-13)
The structure of the gene 5 protein of filamentous bacteriophage Pf1 and its interaction with viral DNA have been investigated by a series of limited proteolysis experiments. The ability of purified proteolytic fragments of the Pf1 gene 5 protein to
Rama Sethuraman et al.
Clinical chemistry, 53(8), 1489-1494 (2007-06-26)
D-Serine is a coagonist for the glycine-binding site of the N-methyl-D-aspartate receptors and has been implicated in various neuropsychiatric functions such as learning, memory, and nociception, as well as schizophrenia and Alzheimer disease. We developed an HPLC method for D-
A Zaramella et al.
Journal of combinatorial chemistry, 3(5), 410-420 (2001-09-11)
The presence of dansyl or dabsyl chromogenic moieties in a solid-phase analytical construct, an assembly of linkers/spacers/sensitizers for improving analytical characterization, allows the accurate estimation of products from solid-phase synthesis by UV detection during liquid chromatography-mass spectrometry analysis in the
P A de Witte et al.
Analytical biochemistry, 226(1), 1-9 (1995-03-20)
We have developed a new method for the quantification of phosphoserine, phosphothreonine, and phosphotyrosine as dabsyl derivatives in acid-hydrolyzed extracts of 32P-labeled A431 cells. In the first step the phosphamino acids are concentrated using a disposable anion-exchange column. Subsequently, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service