Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Biochem/physiol Actions
Simvastatin is a specific inhibitor of HMG-CoA reductase and cholesterol lowering drug.
Simvastatin is a specific inhibitor of HMG-CoA reductase, the enzyme that catalyzes the conversion of HMG-CoA to mevalonate, an early step in cholesterol biosynthesis. It is used in the treatment of hypercholesterolemia, as it reduces levels of low-density lipoproteins and triglycerides, and raises high-density lipoprotein levels. Simvastatin is a lactone that is readily hydrolyzed in vivo to the corresponding β-hydroxyacid, and can be activated prior to use with NaOH in EtOH treatment. It is a synthetic analog of lovastatin (Cat. No. M2147).
Familial hypercholesterolemia is characterized by impaired uptake of cholesterol in peripheral tissues, including the liver and the pancreas. In contrast, statins increase the cellular cholesterol uptake and are associated with increased risk for type 2 diabetes mellitus. We hypothesize that
The Lancet. Neurology, 13(7), 666-675 (2014-05-20)
The benefit of statins in patients with acute aneurysmal subarachnoid haemorrhage is unclear. We aimed to determine whether simvastatin 40 mg could improve the long-term outcome in patients with this disorder. In this international, multicentre, randomised, double-blind trial, we enrolled
The American journal of pathology, 184(8), 2322-2332 (2014-06-08)
Myotonic dystrophy type 2 (DM2) is a multisystemic disorder caused by a (CCTG)n repeat expansion in intron 1 of CNBP. Transcription of the repeats causes a toxic RNA gain of function involving their accumulation in ribonuclear foci. This leads to
Expert opinion on drug delivery, 8(9), 1205-1220 (2011-05-28)
Chronic Obstructive Pulmonary Disease (COPD) is a severe disease that leads to a non-reversible obstruction of the small airways. The prevalence of this disease is rapidly increasing in developed countries, and in 2020 it has been predicted that this disease
In phase 2 studies, evolocumab, a fully human monoclonal antibody to PCSK9, reduced LDL-C levels in patients receiving statin therapy. To evaluate the efficacy and tolerability of evolocumab when used in combination with a moderate- vs high-intensity statin. Phase 3