Skip to Content
Merck
All Photos(1)

Key Documents

36903

Supelco

PCB No 138 solution

10 μg/mL in isooctane, analytical standard

Synonym(s):

2,2′,3,4,4′,5′-Hexachlorobiphenyl solution, 2,2′,3,4,4′,5′-PCB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl6
CAS Number:
Molecular Weight:
360.88
Beilstein:
2508233
Ballschmiter Number:
138
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in isooctane

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

2-8°C

SMILES string

Clc1ccc(c(Cl)c1Cl)-c2cc(Cl)c(Cl)cc2Cl

InChI

1S/C12H4Cl6/c13-7-2-1-5(11(17)12(7)18)6-3-9(15)10(16)4-8(6)14/h1-4H

InChI key

RPUMZMSNLZHIGZ-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

10.4 °F - closed cup

Flash Point(C)

-12 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Gribaldo et al.
Journal of toxicology and environmental health. Part A, 55(2), 121-131 (1998-10-07)
The effects of two substituted polychlorinated biphenyls, the 3,4,5,3',4,5' (PCB-169) and the 2,3,4,2',4',5' (PCB-138) forms, were examined on the expression of c-myc, c-jun, c-ras, and jun-b in 3T3-L1 cells. Northern blot analysis demonstrated that the two PCBs, which exhibit a
Roberta Campagna et al.
Toxicological sciences : an official journal of the Society of Toxicology, 123(1), 170-179 (2011-06-16)
Developmental exposure to polychlorinated biphenyls (PCBs) has been associated with cognitive deficits in humans and laboratory animals by mechanisms that remain unknown. Recently, it has been shown that developmental exposure to 2,2',3,4,4',5'-hexachlorobiphenyl (PCB138), a food-relevant PCB congener, decreases the learning
Linlong Yu et al.
Chemosphere, 90(9), 2347-2351 (2012-11-13)
Sensitized photodechlorination of polychlorinated biphenyl, PCB 138, in three different surfactant solutions was studied. The sensitizer of choice was leuco-methylene blue, which was produced in situ from methylene blue using either triethylamine or sodium borohydride. Three types of surfactants, anionic
Somiranjan Ghosh et al.
Environment international, 37(5), 838-857 (2011-04-08)
Several reports have indicated that low level of polychlorinated biphenyl (PCB) exposure can adversely affect a multitude of physiological disorders and diseases in in vitro, in vivo, and as reported in epidemiological studies. This investigation is focused on the possible
O Roots et al.
Chemosphere, 35(5), 979-985 (1997-09-23)
A comparison of PCB (polychlorinated biphenyls) contents of biota in different areas of the Baltic Sea suggests the northern part of the Gulf of Riga and Väinameri Sea (Estonian coastal water) as reference area for the Baltic Sea. The concentrations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service